Skip to Content
Merck
All Photos(1)

Key Documents

MABS1963

Sigma-Aldrich

Anti-PEG Antibody, clone AGP3

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
€161.00
100 μG
€382.00

€161.00


Estimated to ship on07 April 2025Details


Request a Bulk Order

Select a Size

Change View
25 μG
€161.00
100 μG
€382.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€161.00


Estimated to ship on07 April 2025Details


Request a Bulk Order

antibody form

purified antibody

clone

AGP3
monoclonal

purified by

ion-exchange chromatography

species reactivity

mouse, all, all

concentration

(Please refer to lot specific datasheet.)

technique(s)

ELISA: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgMκ

General description

PEG-conjugated to E.coli beta-glucuronidase.

Specificity

Clone AGP3 is a mouse monoclonal antibody with specificity for polyehtylene glycol backbone.

Application

Western Blotting Analysis: A represenative lot detected PEG in Western Blotting applications (Cheng, T.L., et. al. (2005). Bioconjug Chem. 16(5):1225-31).

ELISA Analysis: A represenative lot detected PEG in ELISA applications (Cheng, T.L., et. al. (2005). Bioconjug Chem. 16(5):1225-31).

Immunohistochemistry Analysis: A represenative lot detected PEG in Immunohistochemistry applications (Ton, N.C., et. al. (2007). Clin Cancer Res. 13(23):7113-8).

Quality

Evaluated by ELISA with immobilized Polyethylene Glycol (10K-PEG, 3K-PEG, and 2K-mPEG).

ELISA Analysis: 20 µg/mL of Polyethylene Glycol (PEG) (10K-PEG, 3K-PEG, and 2K-mPEG) were detected by this antibody.

Physical form

Purified mouse monoclonal antibody IgM in PBS with 0.04% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service