Skip to Content
Merck
All Photos(1)

Key Documents

MABS1906

Sigma-Aldrich

Anti-MULT1 Antibody, clone 1D6

clone 1D6, from rat

Synonym(s):

MULT-1, ULBP1, mouse UL16-binding protein-like Transcript 1, NKG2DL, NKG2D ligand

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
€130.00
100 μG
€353.00

€130.00


Estimated to ship on15 April 2025Details


Request a Bulk Order

Select a Size

Change View
25 μG
€130.00
100 μG
€353.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€130.00


Estimated to ship on15 April 2025Details


Request a Bulk Order

biological source

rat

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1D6, monoclonal

species reactivity

mouse

packaging

antibody small pack of 25 μg

technique(s)

ELISA: suitable
flow cytometry: suitable
immunocytochemistry: suitable

isotype

IgG2aκ

UniProt accession no.

target post-translational modification

unmodified

Gene Information

mouse ... Ulbp1(77777)

Related Categories

General description

UL16-binding protein-like transcript 1 splicing variant (UniProt: Q330P3; also known as MULT1, MULT-1, ULBP1) is encoded by the Ulbp1 gene in murine species. MULT-1 is a MHC Class I-like molecule that serves as a high affinity ligand for natural Killer Group 2D (NKG2D) receptor. It is a type I transmembrane protein that is synthesized with a signal peptide (aa 1-25), which is subsequently cleaved off to generate the mature form. Its transmembrane domain is localized to amino acids 124 to 146 and its extracellular domain contains an alpha 1 and alpha 2 like domain with two intrachain disulfide bonds. MULT-1 expression is reported to be low or absent in normal adult tissues, however, under conditions of stress and in tumor cells its levels are elevated. Many tumor cells are shown to release soluble NKG2D ligands through proteolytic shedding, alternative splicing, or exosome secretion. In mice MULT-1 is shown to cause NK cell activation and tumor rejection. Mice subjected to cytomegalovirus (MCMV) infection display strong expression of Ulbp1 (Mult1) gene, but surface expression of MULT-1 is nevertheless completely abolished by the virus. Fc receptor fcr-1 is reported to down-regulate MULT-1 levels by interfering with surface MULT-1 and causing its subsequent degradation in lysosomes. This effect of Fcr-1 can be completely abolished by imipramine (Cat. No. I0899), which blocks AP2-mediated clathrin pathway. (Ref.: Lenac, T., et al. (2006). J. Exp. Med. 203(8); 1843 1850; Krmpotic, A., et al. (2005). J. Exp. Med. 201(2): 211-220; Deng, W., et al. (2015). Science. 348(6230); 136-139).

Specificity

Clone 1D6 is a rat monoclonal antibody that detects UL16-binding protein-like transcript 1 splicing variant in mouse.

Immunogen

Epitope: extracellular domain
Recombinant fragment corresponding to 186 amino acids from the extracellular domain of murine UL16-binding protein-like transcript 1 splicing variant (MULT-1).

Application

Anti-MULT1, clone 1D6, Cat. No. MABS1906, is a rat monoclonal antibody that detects UL16-binding protein-like transcript 1 splicing variant and has been tested for use in ELISA, Flow Cytometry, and Immunocytochemistry.
Immunocytochemistry Analysis: A representative lot detected MULT1 in Immunocytochemistry applications (Lenac, T., et. al. (2006). J Exp Med. 203(8):1843-50; Krmpotic, A., et. al. (2005). J Exp Med. 201(2):211-20).

Flow Cytometry Analysis: A representative lot detected MULT1 in Flow Cytometry applications (Lenac, T., et. al. (2006). J Exp Med. 203(8):1843-50; Krmpotic, A., et. al. (2005). J Exp Med. 201(2):211-20).

ELISA Analysis: A representative lot detected MULT1 in ELISA applications (Sheppard, S., et. al. (2017). Nat Commun. 8:13930; Krmpotic, A., et. al. (2005). J Exp Med. 201(2):211-20).
Research Category
Signaling

Quality

Evaluated by Flow Cytometry in Splenocytes from BALB/c mouse.

Flow Cytometry Analysis: 1 µg of this antibody detected MULT1 in one million splenocytes from BALB/c mouse.

Target description

27.59 kDa calculated.

Physical form

Format: Purified
Protein G purified
Purified rat monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service