Skip to Content
Merck
All Photos(3)

Key Documents

MABS1905

Sigma-Aldrich

Anti-TMEPAI Antibody, clone 9F10

clone 9F10, from mouse

Synonym(s):

Prostate transmembrane protein androgen induced 1, Solid tumor-associated 1 protein, Transmembrane prostate androgen-induced protein

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€317.00

€317.00


Estimated to ship on07 May 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€317.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€317.00


Estimated to ship on07 May 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

9F10, monoclonal

species reactivity

human, monkey

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... PMEPA1(56937)

General description

Protein TMEPAI (UniProt: Q969W9; also known as Prostate transmembrane protein androgen induced 1, Solid tumor-associated 1 protein, Transmembrane prostate androgen-induced protein) is encoded by the PMEPA1 (also known as STAG1, TMEPAI) gene (Gene ID: 56937) in human. TMEPAI is a type Ib transmembrane protein containing two TMEPAI contains 2 WW-binding motifs and a SMAD interaction motif. The WW-binding motifs mediate its interaction with NEDD4 and the SMAD interaction motif is required for interaction with SMAD2 and SMAD3 and for the negative regulation of TGF-beta signaling. TMEPAI antagonizes TGF-beta signaling by interfering with TGF-beta type I receptor-induced R-SMAD phosphorylation. It competes with SMAD anchor for receptor activation for R-SMAD binding, thereby sequestering R-SMADs from TGF-beta receptor I kinase activation. By acting as a negative regulator of TGF-beta signaling, TMEPAI plays a role in cell proliferation, differentiation, apoptosis, motility, extracellular matrix production, and immunosuppression. Higher expression of TMEPAI is observed in prostate and ovaries and the expression can be up-regulated by androgen and TGF-beta (at protein level).TMEPAI is constitutively and highly expressed in many types of cancer and has been implicated in poor prognosis. TMEPAI is also reported to be involved in down-regulation of the androgen receptor (AR), enhancing ubiquitination and proteasome-mediated degradation of AR. (Ref.: Nguyen THV et al. (2014). Cancer Sci. 105: 334 341; Watanabe, Y et al. (2010). Mol. Cell 37(1): 123-134).

Specificity

Clone 9F10 detects TMEPAI in COS-7 cells overexpressing TEMPAI. It targets an epitope of 39 amino acids from the C-terminal region in isoform 1. Also shows reactivity with isoform 2 that has 100% sequence homology in this epitope region.

Immunogen

GST-tagged recombinant fragment corresponding to 39 amino acids from the C-terminal region of human TMEPAI.

Application

Immunoprecipitation Analysis: A representative lot detected TMEPAI in Immunoprecipitation applications (Vo Nguyen, T.T., et. al. (2014). Cancer Sci. 105(3):334-41).

Western Blotting Analysis: A representative lot detected TMEPAI in lysate from COS-7 cells overexpressing TEMPAI. (Courtesy of Yukihide Watanabe, Ph.D. and Mitsuyasu Kato, Ph.D., University of Tsukuba, Japan).

Western Blotting Analysis: 8 ug/mL from a representative lot detected TMEPAI in TGF-beta stimulated HaCaT cell lysates (Courtesy of Yukihide Watanabe, Ph.D. and Mitsuyasu Kato, Ph.D., University of Tsukuba, Japan).

Immunocytochemistry Analysis: A representative lot detected TMEPAI in Immunocytochemistry applications (Vo Nguyen, T.T., et. al. (2014). Cancer Sci. 105(3):334-41).

Western Blotting Analysis: A representative lot detected TMEPAI in Western Blotting applications (Vo Nguyen, T.T., et. al. (2014). Cancer Sci. 105(3):334-41).
Research Category
Cell Structure
This mouse monoclonal Anti-TMEPAI, Antibody, clone 9F10, Cat. No. MABS1905 is used in Immunocytochemistry, Immunoprecipitation, and Western blotting for the detection of TMEPAI.

Quality

Evaluated by Western Blotting in COS-7 overexpressing TMEPAI lysate.

Western Blotting Analysis: 0.02 µg/mL of this antibody detected TMEPAI in 45 µg of lysate from COS-7 cells overexpressing TMEPAI.

Target description

~32 kDa observed; 31.61 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service