MABS1894
Anti-TAB-2 Antibody, clone 1D5.2
clone 1D5.2, from mouse
Synonym(s):
TGF-beta-activated kinase 1 and MAP3K7-binding protein 2, Mitogen-activated protein kinase kinase kinase 7-interacting protein 2, TAK1-binding protein 2, TAB-2, TGF-beta-activated kinase 1-binding protein 2
About This Item
Recommended Products
biological source
mouse
antibody form
purified antibody
antibody product type
primary antibodies
clone
1D5.2, monoclonal
species reactivity
human
packaging
antibody small pack of 25 μg
technique(s)
western blot: suitable
isotype
IgG2bκ
NCBI accession no.
UniProt accession no.
target post-translational modification
unmodified
Gene Information
human ... TAB2(23118)
Related Categories
General description
Specificity
Immunogen
Application
Quality
Western Blotting Analysis: 1 µg/mL of this antibody detected TAB-2 in 10 µg of TF-1 cell lysate.
Target description
Physical form
Other Notes
Not finding the right product?
Try our Product Selector Tool.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service