MABS1753
Anti-TC PTPase (PTPN2) Antibody, clone 9E9.1
clone 9E9.1, from mouse
Synonym(s):
Tyrosine-protein phosphatase non-receptor type 2, EC: 3.1.3.48, T-cell protein-tyrosine phosphatase, TCPTP
Select a Size
About This Item
Recommended Products
biological source
mouse
Quality Level
antibody form
purified antibody
antibody product type
primary antibodies
clone
9E9.1, monoclonal
species reactivity
human
packaging
antibody small pack of 25 μg
technique(s)
immunohistochemistry: suitable (paraffin)
western blot: suitable
isotype
IgG1κ
NCBI accession no.
UniProt accession no.
shipped in
ambient
target post-translational modification
unmodified
Gene Information
human ... PTPN2(5771)
Related Categories
General description
Specificity
Immunogen
Application
Western Blotting Analysis: 0.5 µg/mL from a representative lot detected TC PTPase (PTPN2) in 10 µg of Jurkat cell lysate.
Signaling
Quality
Western Blotting Analysis: 0.5 µg/mL of this antibody detected TC PTPase (PTPN2) in 10 µg of K562 cell lysate.
Target description
Physical form
Storage and Stability
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service