Skip to Content
Merck
All Photos(1)

Key Documents

MABS165

Sigma-Aldrich

Anti-STAM-binding protein Antibody, clone 4D12.2

clone 4D12.2, from mouse

Synonym(s):

STAMBP, STAM-binding protein, Associated molecule with the SH3 domain of STAM, Endosome-associated ubiquitin isopeptidase, AMSH

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€343.00

€343.00


Estimated to ship on10 March 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€343.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€343.00


Estimated to ship on10 March 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

4D12.2, monoclonal

species reactivity

human

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... STAMBP(10617)

General description

STAM-binding protein (STAMBP) is a critical metalloprotease in ubiquitin-mediated signaling events that specifically cleaves ′Lys-63′-linked polyubiquitin chains but does not cleave ′Lys-48′-linked polyubiquitin chains. STAM-binding protein is a deubiquitinating (DUB) isopeptidase also known as AMSH, for associated molecule with the SH3 domain of STAM. STAM-binding protein plays a role in cell growth and MYC induction, potentiates BMP signaling and is key in regulating the cell surface receptor-mediated endocytosis and ubiquitin-dependent sorting of receptors to lysosomes. STAM-binding protein interacts with other key proteins including STAM1, SMAD6/7, SMURF2, and CHMP3. STAM-binding protein is widely expressed and can be found localized to the nucleus, cytoplasm and endosome. Defects in STAM-binding protein result in a congenital syndrome named Microcephaly-capillary malformation syndrome.

Immunogen

GST-tagged recombinant protein corresponding to Human STAM-binding protein.

Application

Immunocytochemistry analysis: 1:50 dilution from a representative lot detected STAM binding protein in A431 cells
DyLight® is a trademark of Thermo Fisher Scientific, Inc. and its subsidiaries.
Research Category
Signaling
Research Sub Category
Ubiquitin & Ubiquitin Metabolism
This Anti-STAM-binding protein Antibody, clone 4D12.2 is validated for use in Western Blotting and Immunocytochemistry for the detection of STAM-binding protein.

Quality

Routinely evaluated by western blotting on Jurkat cell lysate
Western Blotting Analysis: 1 µg/mL of this antibody detected STAM binding protein in 10 µg of Jurkat cell lysate

Target description

~50 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ supernatant in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Legal Information

DyLight is a registered trademark of Pierce Biotechnology, Inc.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service