Skip to Content
Merck
All Photos(2)

Key Documents

MABS1267

Sigma-Aldrich

Anti-Nuclear Pore Complex Proteins Antibody, clone 414

clone 414, from mouse

Synonym(s):

Nuclear pore glycoprotein p62, 62 kDa nucleoporin, Nucleoporin Nup62, Nuclear pore complex proteins

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€373.00

€373.00


Estimated to ship on22 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€373.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€373.00


Estimated to ship on22 April 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

414, monoclonal

species reactivity

yeast, mouse, rat, human

technique(s)

electron microscopy: suitable
immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... NUP62(23636)

General description

Nuclear pore glycoprotein p62 (UniProt P17955; also known as 62 kDa nucleoporin, Nucleoporin Nup62) is encoded by the Nup62 gene (Gene ID 65274) in rat species. The nuclear pore complex (NPC) acts as a passageway for nucleocytoplasmic transport. Soluble cargo-protein complexes navigate through the pore by binding to phenylalanine-glycine (FG)-repeat proteins attached to the channel walls. The Nup62 complex is located in the center of the NPC and contains the FG-repeat proteins Nup62, Nup54, and Nup58 associated with each other via conserved coiled-coil segments in a 1:1:1 stiochiometric ratio. The nuclear envelope breaks down during prometaphase. The disassembled nucleoporin subcomplexes act as critical regulators of various mitotic events to ensure proper chromosome segregation. Specifically, Nup62 is reported to localize to centrosomes/spindle poles, where it associates with the centrosomal proteins gamma-tubulin and hSAS-6. Nup62 is involved in the recruitment of critical proteins to the centrosome and its depletion results in mislocalization of several centrosomal components.

Specificity

Clone 414 immunostained rat liver cell nuclear rim and yeast cell nuclear envelope (Aris, J.P., and Blobel, G. (1989). J. Cell Biol. 108(6):2059-2067; Davis, L.I., and Blobel, G. (1986). Cell. 45(5):699-709).

Immunogen

Triton X-100 treated rat liver nuclei.

Application

Anti-Nuclear Pore Complex Proteins Antibody, clone 414 is an antibody against Nuclear Pore Complex Proteins for use in Immunocytochemistry, Immunoprecipitation, Western Blotting, Electron Microscopy.
Immunocytochemistry Analysis: A 1:200 dilution from a representative lot immunostained NIH/3T3 cell nuclear rim.
Immunocytochemistry Analysis: A representative lot immunostained yeast nuclear envelope with a punctate and patchy pattern (Aris, J.P., and Blobel, G. (1989). J. Cell Biol. 108(6):2059-2067).
Immunocytochemistry Analysis: Representative lots detected a punctate staining pattern of Nup62 at the nuclear rim by fluorescent immunocytochemistry using 2% formaldehyde-fixed, methanol-permeabilized Buffalo rat liver (BRL) cells (Davis, L.I., and Blobel, G. (1987). Proc. Natl. Acad. Sci. U. S. A. 84(21):7552-755; Davis, L.I., and Blobel, G. (1986). Cell. 45(5):699-709).
Electron Microscopy: A representative lot immunostained yeast nuclear envelope using 3% paraformaldehyde/0.2% glutaraldehyde-fixed yeast nuclei LR White sections (Aris, J.P., and Blobel, G. (1989). J. Cell Biol. 108(6):2059-2067).
Electron Microscopy: A representative lot immunostained the pore complexes in thin sections of isolated rat liver nuclei extracted with 2% Triton X-100 and fixed with 0.05% glutaraldehyde (Davis, L.I., and Blobel, G. (1986). Cell. 45(5):699-709).
Immunoprecipitation Analysis: A representative lot immunoprecipited a ~100 kDa (p110) and a ~95 kDa (p95) protein species from yeast nuclear extract. An additional ~55 kDa protein was immunoprecipitated by clone 414 using yeast cytosolic fraction or whole cell lysate (Aris, J.P., and Blobel, G. (1989). J. Cell Biol. 108(6):2059-2067).
Immunoprecipitation Analysis: A representative lot immunoprecipitated three major GlcNAcylated prtoein species of 62, 175, and 270 kDa from SDS-solubilized pore complex-lamina extract of Buffalo rat liver (BRL) cell nuclei preparation that had been labeled with UDP-[3H]Gal by galactosyltransferase (Davis, L.I., and Blobel, G. (1987). Proc. Natl. Acad. Sci. U. S. A. 84(21):7552-755).
Immunoprecipitation Analysis: A representative lot immunoprecipitated GlcNAcylated 62 kDa protein (p62) from the SDS-solubilized pore complex-lamina extract, as well as a less glycosylated p61 cytoplasmic form from the postmitochondrial supernatant of Buffalo rat liver (BRL) cells (7552-755; Davis, L.I., and Blobel, G. (1986). Cell. 45(5):699-709).
Western Blotting Analysis: A representative lot detected a ~100 kDa (p110) and a ~95 kDa (p95) immunoreactive bands in yeast nuclear extract (Aris, J.P., and Blobel, G. (1989). J. Cell Biol. 108(6):2059-2067).
Western Blotting Analysis: A representative lot detected a ~62 kDa (p62) and a ~200 kDa target bands associated with nuclear pore complex-lamina of rat liver nuclei preparation even following sequential nucleaases, Triton X-100, and 140 mM NaCl treatments (Davis, L.I., and Blobel, G. (1986). Cell. 45(5):699-709).
Research Category
Signaling
Research Sub Category
Chromatin Biology

Quality

Evaluated by Immunocytochemistry in HeLa cells.

Immunocytochemistry Analysis: A 1:200 dilution of this antibody immunostained HeLa cell nuclear rim.

Target description

53.40 kDa (Rat Nup62) calculated. ~62/175/270 kDa (Rat liver nuclear extract) and ~95/100 kDa (Yeast nuclear extract) reported (Aris, J.P., and Blobel, G. (1989). J. Cell Biol. 108(6):2059-2067; Davis, L.I., and Blobel, G. (1986). Cell. 45(5):699-709). Uncharacterized band(s) may appear in some lysates.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael Sean Mauro et al.
eLife, 11 (2022-07-20)
The nuclear envelope (NE) assembles and grows from bilayer lipids produced at the endoplasmic reticulum (ER). How ER membrane incorporation coordinates with assembly of nuclear pore complexes (NPCs) to generate a functional NE is not well understood. Here, we use

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service