Skip to Content
Merck
All Photos(1)

Key Documents

MABS1250

Sigma-Aldrich

Anti-Mineralocorticoid Receptor Antibody, clone 2D6

clone 2D6, from mouse

Synonym(s):

MR, Nuclear receptor subfamily 3 group C member 2, Mineralocorticoid Receptor

Sign Into View Organizational & Contract Pricing

Select a Size

200 μG
€429.00

€429.00


Estimated to ship on11 April 2025Details


Request a Bulk Order

Select a Size

Change View
200 μG
€429.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

€429.00


Estimated to ship on11 April 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2D6, monoclonal

species reactivity

rat

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

General description

Mineralocorticoid receptor (UniProt P22199; also known as MR, Nuclear receptor subfamily 3 group C member 2) is encoded by the Nr3c2 (also known as MCR, Mlr) gene (Gene ID 25672) in rats. Mineralocorticoid receptor (MR) is expressed in multiple tissues, including renal epithelia, smooth muscle, endothelium, cardiomyocytes, and hippocampal neurons, where it mediates diverse functions. MR is activated by aldosterone and also by cortisol in cells that do not express 11β-hydroxysteroid dehydrogenase type 2 (11βHSD2). MR is normally activated by aldosterone, which is produced by adrenal glomerulosa in response to intravascular volume depletion and hyperkalemia. Consistently, gain of function mutations in MR lead to severe hypertension, often with hypokalemia, while loss of function mutations result in neonatal hypotension. Ser843 phosphorylation in the MR ligand-binding domain is reported to prevent MR ligand binding and activation. MR pS843 is found exclusively in intercalated cells of the distal nephrons in the kidney. Intravascular volume depletion, angiotensin II and WNK4 signaling reduce MR pS843 levels, whereas hyperkalemia increases MR pS843.

Specificity

Epitope is present in all 3 alternative spliced isoforms

Immunogen

Conjugated linear peptide corresponding to the N-terminus of rat Mineralocorticoid Receptor.
Epitope: Near N-terminus

Application

Detect Mineralocorticoid Receptor using this Anti-Mineralocorticoid Receptor Antibody, clone 2D6 validated for use in Western Blotting, Immunohistochemistry and Immunocytochemistry.
Immunohistochemistry Analysis: A representative lot detected Mineralocorticoid Receptor in hippocampus, dentate gyrate, choroid plexus, heart, and rat kidney (Gomez-Sanchez, C.E., et al. (2006). Endocrinology. 147(3):1343-1348).
Immunocytochemistry Analysis: A representative lot detected Mineralocorticoid Receptor in mouse kidney (Shibata, S., et al. (2013). Cell Metabolism. 18:660-671).
Western Blotting Analysis: A representative lot detected endogenous Mineralocorticoid Receptor (MR) in rat hippocampal cytosolic preparation and exogenously expressed EGFP-rat MR fusion protein in CHO cells (Gomez-Sanchez, C.E., et al. (2006). Endocrinology. 147(3):1343-1348).
Western Blotting Analysis: A representative lot detected endogenous Mineralocorticoid Receptor (MR) in mouse kidney cytosolic fraction and exogenously expressed human MR in COS-7 cells (Shibata, S., et al. (2013) Cell Metab. 18(5):660-671)
Research Category
Signaling
Research Sub Category
Signaling Neuroscience

Quality

Evaluated by Western Blotting in rat brain cytosol tissue lysate.

Western Blotting Analysis: A 1:500 dilution of this antibody detected Mineralocorticoid Receptor in 10 µg of rat brain cytosol tissue lysate.

Target description

~107 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2aκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service