Skip to Content
Merck
All Photos(2)

Key Documents

MABS1248

Sigma-Aldrich

Anti-PPIase FKBP4 Antibody, clone KN382/EC1

clone KN382/EC1, from mouse

Synonym(s):

Peptidyl-prolyl cis-trans isomerase FKBP4, PPIase FKBP4, 52 kDa FK506-binding protein, 52 kDa FKBP, FKBP-52, 59 kDa immunophilin, p59, FK506-binding protein 4, FKBP-4, FKBP59, HSP-binding immunophilin, HBI, Immunophilin FKBP52

Rotamase, Peptidyl

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€429.00

€429.00


Estimated to ship on14 March 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€429.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€429.00


Estimated to ship on14 March 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

KN382/EC1, monoclonal

species reactivity

human, rabbit

should not react with

mouse, rat

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
radioimmunoassay: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... FKBP4(2288)

General description

Peptidyl-prolyl cis-trans isomerase FKBP4 (EC 5.2.1.8; UniProt P27124; also known as 52 kDa FK506-binding protein, 52 kDa FKBP, 52 kDa immunophilin, FK506-binding protein 4, FKBP-4, FKBP-52, FKBP59, HBI, HSP-binding immunophilin, Immunophilin FKBP52, PPIase FKBP4, p59, Rotamase) is encoded by the FKBP4 (also known as P59) gene (Gene ID 100009148) in rabbit. PPIase FKBP4 (FKBP52, FKBP59, p59) is an immunophilin that binds the immunosuppressant drugs FK506 and rapamycin. FKBP59 was originally identified in the rabbit as a component of the unliganded, nontransformed steroid hormone receptor oligomeric complexes, where it was found associated with the chaperon protein hsp90 involved in protein folding. All the immunophilins are implicated in protein conformation regulations by catalyzing the cis-trans isomerization of proline imidic peptide bonds via their peptidylprolyl isomerase (rotamase) activity. FKBP59 is expressed at a higher level in the testis than in other organs. A high FKBP59 expression occurs in germ cells that have reached the same developmental stages either during prepubertal development or differentiation along the seminiferous tubules in the adult. The developmental stage-specific expression of FKBP59 during male germ cell differentiation suggests a role during the cell division.

Immunogen

Nonactivated (8.5S) rabbit uterine progestin receptor.

Application

This Anti-PPIase FKBP4 Antibody, clone KN382/EC1 is validated for use in Western Blotting, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation and Radioimmunoassay for the detection of PPIase FKBP4.
Western Blotting Analysis: 0.5 µg/mL from a representative lot detected PPIase FKBP4 in 10 µg of human brain tissue lysate.
Immunocytochemistry Analysis: A representative lot detected predominantly cytosolic immunoreactivity in human JAR choriocarcinoma cells by fluorescent immunocytochemistry (Sanchez, E.R., et al. (1990). Biochemistry. 29(21):5145-5152).
Immunohistochemistry Analysis: A representative lot stained both the cytosol and nucleus of epithelial and stromal cells using frozen uterus sections from estradiol-stimulated rabbits, while predominant nuclear staining was seen when using paraffin-embedded sections (Gasc, J.M., et al. (1990). Exp. Cell Res. 186(2):362-367).
Immunohistochemistry Analysis: A representative lot detected predominantly cytoplasmic PPIase FKBP4 (FKBP59) immunoreactivity in the early spermatids (eS), while both cytosolic and nuclear staining was seen in the midpachytene spermatocytes using paraformaldehyde-fixed, paraplast-embedded rabbit testis sections corresponding to stage VIII of the seminiferous epithelium cycle (Sananes, N., et al. (1998). Biol. Reprod. 58(2):353-360).
Immunoprecipitation Analysis: A representative lot co-immunoprecipitated Hsp90 with p59 from human lymphocyte IM-9 cytosolic extracts (Sanchez, E.R., et al. (1990). Biochemistry. 29(21):5145-5152).
Immunoprecipitation Analysis: A representative lot detected heat shock treatment-induced p59 (a.k.a. p56) upregulation in human IM-9 lymphocytes by IP (Sanchez, E.R. (1990). J Biol Chem. 265(36):22067-22070).
Radioimmunoassay Analysis: A representative lot immunoprecipitated [3H] FK506-binding activity in the cytosolic preparations from human Jurkat and IM-9, but not mouse S49 lymphocytes (Tai, P.K., et al. (1992). Science. 256(5061):1315-1318).
Western Blotting Analysis: A representative lot detected the presence of p59 (FKBP59) in the FK506 pull-down from human lymphocyte IM-9 cytosolic preparations (Tai, P.K., et al. (1992). Science. 256(5061):1315-1318).
Western Blotting Analysis: A representative lot detected p59 in both the cytosolic and nuclear fractions of uterus extracts from estradiol-stimulated rabbits (Gasc, J.M., et al. (1990). Exp. Cell Res. 186(2):362-367).

Quality

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected PPIase FKBP4 in 10 µg of HeLa cell lysate.

Target description

~59 kDa observed

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service