Skip to Content
Merck
All Photos(3)

Key Documents

MABS1229

Sigma-Aldrich

Anti-SKP1 Antibody, clone 3F3.1

clone 3F3.1, from mouse

Synonym(s):

S-phase kinase-associated protein 1, Cyclin-A/CDK2-associated protein p19, OCP-2, OCP-II, Organ of Corti protein 2, Organ of Corti protein II, p19A, p19skp1, RNA polymerase II elongation factor-like protein, SIII, Transcription elongation factor B polype

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€280.00

€280.00


Estimated to ship on26 March 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€280.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€280.00


Estimated to ship on26 March 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3F3.1, monoclonal

species reactivity

mouse, human

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SKP1(6500)

General description

S-phase kinase-associated protein 1 (UniProt P63208; also known as Cyclin-A/CDK2-associated protein p19, OCP-2, OCP-II, Organ of Corti protein 2, Organ of Corti protein II, p19A, p19skp1, RNA polymerase II elongation factor-like protein, SIII, Transcription elongation factor B polypeptide 1-like) is encoded by the SKP1 (also known as EMC19, OCP2, SKP1A, TCEB1L) gene (Gene ID 6500) in human. Polyubiquitination of proteins is mediated by ubiquitin-activating enzymes (E1s), ubiquitin-conjugating enzymes (E2s), and ubiquitin ligases (E3s). There exist RING finger-type, HECT-type, and U-box type of E3 ligases. Skp1 is one of the three core components of the SCF complex (Skp1, Cullin, F-box) and functions as an adapter that bridges the F-box protein and Cullin (CUL1). SCF complex depends on various F-box proteins for specific recognition and recruitment of substrates. SCF ligase plays an important role in regulating the transitions between G1/S and G2/M phases.

Specificity

Expected to react with both spliced isoforms of human SKP1 reported by UniProt (P63208).

Immunogen

Epitope: Internal.
GST-tagged recombinant human SKP1 internal fragment.

Application

Anti-SKP1 Antibody, clone 3F3.1 is an antibody against SKP1 for use in Western Blotting, Immunocytochemistry.
Immunocytochemistry Analysis: A 1:200 dilution from a representative lot detected SKP1 in HUVECs, A431 and HeLa cells.
Research Category
Signaling
Research Sub Category
Ubiquitin & Ubiquitin Metabolism

Quality

Evaluated by Western Blotting in NIH/3T3 cell lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected SKP1 in 10 µg of NIH/3T3 cell lysate.

Target description

~18 kDa observed. 18.66 kDa (Isoform 1) and 18.06 kDa (Isoform 2) calculated.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2aκ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service