Skip to Content
Merck
All Photos(2)

Key Documents

MABS1198

Sigma-Aldrich

Anti-SPINK1 Antibody, clone 4D11.1

clone 4D11.1, from mouse

Synonym(s):

Serine protease inhibitor Kazal-type 1, Pancreatic secretory trypsin inhibitor, TATI, Tumor-associated trypsin inhibitor

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€330.00

€330.00


Estimated to ship on04 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μL
€330.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€330.00


Estimated to ship on04 April 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4D11.1, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... SPINK1(6690)

General description

Serine protease inhibitor Kazal-type 1 (UniProt P00995; also known as Pancreatic secretory trypsin inhibitor, TATI, Tumor-associated trypsin inhibitor) is encoded by the SPINK1 (also known as PCTT, PSTI, TCP) gene (Gene ID 6690) in human. Serine protease inhibitor Kazal-type 1 encoded by the human SPINK1 gene functions as an acute-phase protein (APP) in the pancreas. Produced initially in a 79-a.a. proform, the signal peptide (a.a. 1-23) is removed posttranslationally to yield the 56-a.a. (a.a. 24-79) mature protein. SPINK1 protein blocks premature activation of trypsinogen (inactive form of trypsin), thereby ensuring the integrity of acinar cells to prevent auto-digestion of the pancreas. SPINK1 is also reported to play a role in tissue repair, apoptosis, reproduction, and behave as a growth factor in other human organs. In addition, SPINK1 protein is known as tumor-associated trypsin inhibitor or TATI and current research activities are focused on the association of various cancers to deregulated SPINK1 activity.

Specificity

Expected to react with both pro- and mature human SPINK1.

Immunogen

GST-tagged recombinant human SPINK1.

Application

Anti-SPINK1, clone 4D11.1, Cat. No. MABS1198, is a highly specific mouse monoclonal antibody that targets SPINK1 and has been tested in Immunohistochemistry and Western Blotting.
Immunohistochemistry Analysis: A 1:250 dilution from a representative lot detected SPINK1 in human pancreas tissue sections.
Research Category
Signaling

Quality

Evaluated by Western Blotting of SPINK1 GST fusion protein.

Western Blotting Analysis (WB): A 1:5000 dilution of this antibody detected 50 ng of recombinant human SPINK1 GST fusion protein.

Target description

6.247/8.507 kDa (mature/pro-form) calculated. Uncharacterized band(s) may appear in some lysates.

Physical form

Format: Purified
Protein G purified.
Purified mouse monoclonal IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4) 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service