Skip to Content
Merck
All Photos(1)

Key Documents

MABS1195

Sigma-Aldrich

Anti-SSH1 Antibody, clone 12C6.1

clone 12C6.1, from mouse

Synonym(s):

Protein phosphatase Slingshot homolog 1, hSSH-1L, Slingshot homolog 1, SSH-1L, SSH-like protein 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€330.00

€330.00


Estimated to ship on04 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€330.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€330.00


Estimated to ship on04 April 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

12C6.1, monoclonal

species reactivity

human

technique(s)

western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SSH1(54434)

General description

Protein phosphatase Slingshot homolog 1 (EC 3.1.3.16; EC 3.1.3.48; UniProt Q8WYL5; also known as hSSH-1L, Slingshot homolog 1, SSH-1L, SSH-like protein 1) is encoded by the SSH1 (also known as KIAA1298, SSH1L) gene (Gene ID 54434) in human. SSH1 is the founding member of the Slingshot family of protein phosphatases that function to destabilize the actin cytoskeleton by reversing cofilin Ser3 phosphorylation and thereby re-activating cofilin. Cofilin plays a central role in regulating actin filament turnover and actin cytoskeletal remodelling by severing and depolymerizing actin filaments. Cofilin activity is negatively regulated by LIM kinases-catalyzed Ser3 phosphorylation. The Slingshot family members include SSH-1L, SSH-2L, SSH-3L, and chronophin. The members differ in their subcellular distribution, F-actin-binding activity, and phosphatase activity, implying related but member-specific functions.

Specificity

Expected to react with spliced isoform 1 (isoform L), isoform 2 (isoform S), isoform 4, and isoform 5, but not isoform 3 (isoform B) reported by UniProt (Q8WYL5).

Immunogen

GST-tagged recombinant protein corresponding to an internal sequence of human SSH1.

Application

Anti-SSH1 Antibody, clone 12C6.1 is an antibody against SSH1 for use in Western Blotting.

Quality

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected SSH1 in 10 µg of HeLa cell lysate.

Target description

~150 kDa observed. 115.5 kDa (isoform 1), 77.43 kDa (isoform 2), 80.68 kDa (isoform 4), and 78.98 kDa (isoform 5) calculated.

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service