Skip to Content
Merck
All Photos(1)

Key Documents

MABS1187

Sigma-Aldrich

Anti-phospho-GADS (Thr262) Antibody, clone 6E11.2

clone 6E11.2, from mouse

Synonym(s):

GRB2-related adapter protein 2, Thr262 phosphorylated, Adapter protein GRID, Thr262 phosphorylated, GRAP-2, Thr262 phosphorylated, GRB-2-like protein, Thr262 phosphorylated, Grb2-related adapter protein downstream of Shc, Thr262 phosphorylated, GRB2L, Th

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€401.00

€401.00


Estimated to ship on22 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€401.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€401.00


Estimated to ship on22 April 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

6E11.2, monoclonal

species reactivity

human

technique(s)

western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pThr262)

Gene Information

human ... GRAP2(9402)

General description

GRB2-related adapter protein 2 (UniProt O75791; also known as Adapter protein GRID, GRAP-2, GRB-2-like protein, Grb2-related adapter protein downstream of Shc, GRB2L, GRBLG, GRBX, Grf-40, Grf40 adapter protein, Growth factor receptor-binding protein, Hematopoietic cell-associated adapter protein GrpL, P38, Protein GADS, SH3-SH2-SH3 adapter Mona) is encoded by the GRAP2 (also known as GADS, GRB2L, GRID) gene (Gene ID 9402) in human. GADS (Grb2-related adapter protein downstream of Shc), GRB2 and GRAP constitute the GRB2 family of adapter proteins characterized by a central SH2 domain (a.a. 58-149 of GADS) flanked between two SH3 domains (a.a. 1-56, 271-330 of GADS). Although the SH2 domains of all three members have similar binding specificities, the C-terminal SH3 domain of GADS possesses unique binding specificity that allows for its interaction with the adapter protein SLP-76 (SH2 domain-containing leukocyte protein of 76 kDa). The GADS-SLP-76 complex is recruited to tyrosine-phosphorylated LAT (linker for activation of T cells) via GADS SH2 domain, allowing the assembly of signaling complexes critical to T-cell activation. In addition, evidences indicate that GADS plays a critical role in the development of BCR-ABL-mediated lymphoid disease. The oncogenic Bcr-Abl fusion protein is shown to bind GADS in a phosphotyrosine-dependent manner and GADS is expressed in a subset of CML cell lines and B-ALL patient samples, where it associates with both BCR-ABL and SLP-76.

Specificity

Clone 6E11.2 detects GADS phosphorylated on Thr262. The corresponding phosphorylation site in human GADS spliced isoform 2 is Thr149 (UniProt O75791-2).

Immunogen

Epitope: pThr262.
KLH-conjugated linear peptide corresponding to a human GADS sequence containing phosphorylated Thr262.

Application

Anti-phospho-GADS (Thr262) Antibody, clone 6E11.2 is an antibody against phospho-GADS for use in Western Blotting.
Research Category
Signaling
Research Sub Category
Immunological Signaling

Quality

Evaluated by Western Blotting in Jurkat cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected GADS Thr262 phosphorylation induction in 10 µg lysate from Calyculin A- and TNFα-treated Jurkat cells.

Target description

~40 kDa observed. 37.91 kDa (isoform 1) and 24.38 kDa (isoform 2) calculated. Uncharacterized band(s) may appear in some lysates.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service