Skip to Content
Merck
All Photos(1)

Key Documents

MABN92-AF647

Sigma-Aldrich

Anti-Iba1/AIF1 Antibody, clone 20A12.1, Alexa Fluor 647 Conjugate

clone 20A12.1, from mouse, ALEXA FLUOR 647

Synonym(s):

Allograft inflammatory factor 1, AIF-1, Daintain, Interferon gamma responsive transcript, Ionized calcium-binding adapter molecule 1, Protein G1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€484.00

€484.00


Available to ship on16 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μL
€484.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.45

€484.00


Available to ship on16 April 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

conjugate

ALEXA FLUOR 647

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

20A12.1, monoclonal

species reactivity

mouse, human, rat

technique(s)

immunofluorescence: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... AIF1(199)

General description

Allograft inflammatory factor 1 (UniProt P55008; also known as AIF-1, Daintain, Interferon gamma responsive transcript, Ionized calcium-binding adapter molecule 1, Protein G1) is encoded by the AIF1 (also known as G1, IBA1) gene (Gene ID 199) in human. Iba1/AIF1 is a 17 kDa interferon-gamma- (IFN-gamma-) inducible, Ca(2+)-binding EF-hand protein. Iba1/AIF1 expression is mostly limited to the macrophage/monocyte lineage, and is augmented by cytokines, such as IFN-gamma. It is reported to be involved in neuronal function, inflammatory responses, allograft rejection, autoimmunity, and macrophage activation/function. It is also shown to be expressed in microglial cells in autominnume disesase models such as experimental autoimmune encephalomyelitis, neuritis and uveitis.

Application

Anti-Iba1/AIF1 Antibody, clone 20A12.1, Alexa Fluor 647 Conjugate is an antibody against AIF1 for use in Immunofluorescence.
The unconjugated antibody (Cat. No. MABN92) is shown to be suitable also for Western blotting and immunohistochemistry applications.

Quality

Evaluated by Immunofluorescence in human spleen tissue.

Immunofluorescence Analysis: A 1:50 dilution of this antibody detected Iba1/AIF1 in human spleen tissue.

Target description

~17 kDa observed.

Other Notes

Concentration: Please refer to lot specific datasheet.

Legal Information

ALEXA FLUOR is a trademark of Life Technologies

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Youngmi Kim et al.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc, 36(1), 100034-100034 (2023-02-15)
Glioblastoma is a heterogeneous tumor for which effective treatment options are limited and often insufficient. Few studies have examined the intratumoral transcriptional and proteomic heterogeneity of the glioblastoma microenvironment to characterize the spatial distribution of potential molecular and cellular therapeutic
Jamie M Walker et al.
Acta neuropathologica communications, 10(1), 23-23 (2022-02-16)
Clinical symptoms correlate with underlying neurodegenerative changes in the vast majority of people. However, an intriguing group of individuals demonstrate neuropathologic changes consistent with Alzheimer disease (AD) yet remain cognitively normal (termed "resilient"). Previous studies have reported less overall neuronal
Fumio Takahashi et al.
iScience, 25(5), 104278-104278 (2022-05-17)
Neurodegeneration is a process involving both cell autonomous and non-cell autonomous neuron loss, followed by a collapse of neural networks, but its pathogenesis is poorly understood. We have previously demonstrated that Eomes-positive helper T (Eomes + Th) cells recognizing LINE-1(L1)-derived prototypic
Emmet Huang-Hobbs et al.
Nature, 619(7971), 844-850 (2023-06-29)
The tumour microenvironment plays an essential role in malignancy, and neurons have emerged as a key component of the tumour microenvironment that promotes tumourigenesis across a host of cancers1,2. Recent studies on glioblastoma (GBM) highlight bidirectional signalling between tumours and
Pei Song et al.
Journal of healthcare engineering, 2021, 3048383-3048383 (2021-10-12)
Cardiopulmonary bypass (CPB) is mainly used during cardiac surgeries that treat ischemic, valvular, or congenital heart disease and aortic dissections. The disorders of central nervous system (CNS) that occur after cardiopulmonary bypass are attracting considerable interest. Postoperative neurocognitive disorders (PND)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service