Skip to Content
Merck
All Photos(3)

Key Documents

MABN898

Sigma-Aldrich

Anti-FFAR3/GPR41 Antibody, clone 16F4.1

clone 16F4.1, from mouse

Synonym(s):

Free fatty acid receptor 3, G-protein coupled receptor 41

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€373.00

€373.00


Estimated to ship on22 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€373.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€373.00


Estimated to ship on22 April 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

16F4.1, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... FFAR3(2865)

General description

Free fatty acid receptor 3 (UniProt O14843; also known as G-protein coupled receptor 41) is encoded by the FFAR3 (also known as GPR41) gene (Gene ID 2865) in human. Free fatty acids (FFAs) act as both energy sources and signaling molecules via G-protein-coupled receptors (GPCRs) called FFARs (FFA receptors). FFAs are categorized by the length of their carbon chains into short-chain FAs (SCFAs; less than 6 carbons), medium-chain FAs (MCFAs; 6–12 carbons), and long-chain FAs (LCFAs; more than 12 carbons). MCFAs and LCFAs activate FFAR1 and FFAR4, whereas SCFAs activate FFAR3 and FFAR2. FFAR3 is an SCFA receptor that can be activated by propionate (C3), butyrate (C4), and valerate (C5). FFAR3 mediates PYY and GLP-1 secretion from endocrine and sympathetic outflow-dependent heart rate regulation and energy expenditure, while β-Hydroxybutyrate, a ketone body produced in the liver during starvation, decreases sympathetic outflow by antagonizing FFAR3. FFAR3 is shown to couple Gαi/o pathway signaling upon activation by propionate binding in primary murine pancreatic beta cells, resulting in an inhibition of glucose-dependent insulin secretion. FFAR3 is a 7-transmembrane (a.a. 20-40, 48-68, 89-111, 133-153, 179-199, 223-243, 259-279) GPCR, having a short N-terminal extracellular portion (a.a. 1-19) and a long C-terminal cytomplasmic tail (a.a. 280-346).

Specificity

Clone 16F4.1 targets an epitope in the C-terminal cytoplasmic domain of human FFAR3/GPR41.

Immunogen

KLH-conjugated linear peptide corresponding to a C-terminal cytoplasmic domain sequence of human FFAR3/GPR41.

Application

Immunohistochemistry Analysis: A 1:250 dilution from a representative lot detected FFAR3/GPR41 in human breast cancer tissue, as well as in normal liver, pancreas, and small intestine tissue sections.
This Anti-FFAR3 Antibody, clone 16F4.1 is validated for use in Western Blotting, Immunohistochemistry (Paraffin) for the detection of GPR41.

Quality

Evaluated by Western Blotting in human adipose tissue lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected FFAR3/GPR41 in 10 µg of human adipose tissue lysate.

Target description

~41 kDa observed. 38.65 kDa calculated.

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Suqin Yang et al.
Natural products and bioprospecting, 14(1), 44-44 (2024-08-12)
Xiaoyankangjun tablet (XYKJP) is a traditional Chinese medicine formulation used to treat intestinal disorders in clinical practice. However, the specific therapeutic mechanism of action of XYKJP in colitis has not yet been elucidated. This study aimed to reveal the multifaceted

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service