Skip to Content
Merck
All Photos(2)

Key Documents

MABN893

Sigma-Aldrich

Anti-ABCA1 Antibody, clone MABI98-7

clone MAB198-7, from rat

Synonym(s):

ATP-binding cassette sub-family A member 1, ATP-binding cassette transporter 1, ATP-binding cassette 1, ABCA 1, ABCA-1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€377.00

€377.00


Estimated to ship on02 May 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€377.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€377.00


Estimated to ship on02 May 2025Details


Request a Bulk Order

biological source

rat

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

MAB198-7, monoclonal

species reactivity

mouse, human

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

bovine ... Abca1(535379)

General description

ATP-binding cassette sub-family A member 1, also known as ATP-binding cassette transporter 1 (ABC-1), or ATP-binding cassette 1, and encoded by the gene name Abca1 or Abc1, is a cAMP-dependent and sulfonylurea-sensitive anion transporter. ABCA1 is a member of the ABC transporter superfamily and the ABCA family. ABCA1 is a key gatekeeper influencing intracellular cholesterol transport. ABCA1 has two homologous halves, each containing a hydrophobic membrane-anchoring domain and an ATP binding cassette (ABC) domain, and may act as a multifunctional polypeptide. ABCA1 has been shown to be down-regulated by endotoxins (LPS) or cytokines (TNF and IL-1) in J774 macrophages. The down-regulation by endotoxin in macrophages is not likely to be mediated by the liver X receptor/retinoic X receptor (LXR/RXR). ABCA1 interacts with MEGF10 and is widely expressed in adult tissues, with the highest levels found in uterus and pregnant uterus. Mutations in the ABCA1 gene are associated with Tangier disease (TD). TD is an autosomal recessive disorder results from an absence of plasma HDL, cholesterol ester depositing in the reticulo-endothelial system and disorders in cellular lipid trafficking.

Immunogen

Epitope: Immunogen is near C-terminal.
Linear peptide corresponding to the C-terminus of Mouse ABCA1.

Application

Research Category
Neuroscience
Research Sub Category
Ion Channels & Transporters
This Anti-ABCA1 antibody is validated for use in WB, IH for the detection of ABCA1.
Western Blotting Analysis: A representative lot of MABN893 deteted ABCA1 in lysate of WT mouse brain and demonstrated a loss of signal in lysates of ABCA1 knock out adult mouse brain (Hu et al., 2008. J Lipid Res. 2008 Feb;49(2):386-93.)
Immunohistochemistry Analysis: A 1:250 dilution from a representative lot detected ABCA1 in human cerebral cortex, thalamus, and kidney tissues.
Produced in collaboration with MAB Institute, Inc.

Quality

Evaluated by Western Blotting in Calyculin and Okadaic Acid treated NIH-3T3 cell lysate.

Western Blotting Analysis: A 1 µg/mL diultion of this antibody detected ABCA1 in 10 µg of Calyculin and Okadaic Acid treated NIH-3T3 cell lysate.

Target description

~254 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified rat monoclonal IgG2aκ in buffer containing PBS with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service