Skip to Content
Merck
All Photos(2)

Key Documents

MABN856

Sigma-Aldrich

Anti-Neurophysin 2/NP-AVP Antibody, clone PS 45

clone PS 45, from mouse

Synonym(s):

Vasopressin-neurophysin 2-copeptin, AVP-NPII, Neurophysin 2, Neurophysin-II

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€395.00

€395.00


Estimated to ship on23 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€395.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€395.00


Estimated to ship on23 April 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

PS 45, monoclonal

species reactivity

mouse, human, bovine, frog, rat

technique(s)

ELISA: suitable
electron microscopy: suitable
immunohistochemistry: suitable (paraffin)
immunoprecipitation (IP): suitable
radioimmunoassay: suitable
western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

rat ... Avp(24221)

General description

The neurophysin 1 and 2 (NPI and NPII) are carriers of peptide hormons oxytocin (OT) and vasopressin (AVP), respectively. Each NP and its associated hormon peptide are synthesized as one precursor protein, OT-NPI (UniProt P01179) encoded by the Oxt (or Ot) gene or AVP-NPII (UniProt P01186) encoded by the Avp gene in rat species. Posttranslational cleavages remove the signal peptide and process the precursor into mature OT and NPI (aa 20-28 and aa 32-135, respectively, of OT-NPI), or AVP and NPII (aa 24-32 and aa 36-128, respectively, of AVP-NPII). OT is a potent hormone and neurotransmitter secreted within the pituitary gland. OT exerts its biological activity through its receptor OXTR (also known as OTR). OT is used clinically for labor induction and may exhibit therapeutic potential for treating social anxiety disorders. AVP functions primarily as an antidiuretic and regulates the concentration of water, glucose, and salts in blood. AVP is secreted from the posterior pituitary, but exerts its biological activity in the kidney via the G protein-coupled receptor AVPR2 (V2R).

Specificity

Clone PS 45 reacts with Vasopressin-neurophysin 2-copeptin precursor, neurophysin 2, but not Arg-vasopressin, copeptin, oxytocin, or neurophysin 1.

Immunogen

Epitope: Near Neurophysin 2 C-terminus

Application

Immunohistochemistry Analysis: A 1:250-1,000 dilution from a representative lot detected Neurophysin 2/NP-AVP in rat brain and rat pituitary tissue.
Radioimmunoassay (RIA): Target specificity of a representative lot was confirmed by RIA. Only rat neurophysin 2, but not neurophysin 1, oxytocin, or angiotensin II, can compete against 125I-neurophysin 2 for binding mAb PS 45 (Ben-Barak, Y., et al. (1985). J Neurosci. 5(1):81-97).
Electron Microscopy (EM): A representative lot immunostained granulated neurosecretory vesicles (NSVs) in all axons, Herring bodies, and terminals by EM using rat posterior pituitary LR White sections (Ben-Barak, Y., et al. (1985). J Neurosci. 5(1):81-97).
Immunoprecipitation Analysis: A representative lot immunoprecipitated radiolabelled neurophysin 2, but not neurophysin 1 (Ben-Barak, Y., et al. (1985). J Neurosci. 5(1):81-97).
ELISA Analysis: A representative lot detected neurophysin 2 immunoreactivity in rat neurophysin preparations by non-sandwich ELISA using either 125I-labelled or HRP-conjugated secondary antibody (Ben-Barak, Y., et al. (1985). J Neurosci. 5(1):81-97).
3. References/ Please add reference:
Ben-Barak, Y., et al. (1985). J Neurosci. 5(1):81-97
This Anti-Neurophysin 2/NP-AVP Antibody, clone PS 45 is validated for use in Western Blotting, Immunohistochemistry (Paraffin), Radioimmunoassay, Electron Microscopy, Immunoprecipitation, ELISA for the detection of Neurophysin 2/NP-AVP.

Quality

Evaluated by Western Blotting in rat pituitary tissue lysate.

Western Blotting Analysis: 2.0 µg/mL of this antibody detected Neurophysin 2/NP-AVP in 10 µg of rat pituitary tissue lysate.

Target description

~15 kDa observed

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Iwona Olejniczak et al.
Frontiers in physiology, 13, 909795-909795 (2022-10-25)
Searching for food follows a well-organized decision process in mammals to take up food only if necessary. Moreover, scavenging is preferred during their activity phase. Various time-dependent regulatory processes have been identified originating from the suprachiasmatic nuclei (SCN), which convert
Juan Pablo Arroyo et al.
JCI insight, 7(24) (2022-11-04)
Vasopressin has traditionally been thought to be produced by the neurohypophyseal system and then released into the circulation where it regulates water homeostasis. The questions of whether vasopressin could be produced outside of the brain and if the kidney could
Stefano Musardo et al.
eLife, 11 (2022-04-23)
Social interaction during adolescence strongly influences brain function and behavior, and the recent pandemic has emphasized the devastating effect of social distancing on mental health. While accumulating evidence has shown the importance of the reward system in encoding specific aspects

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service