Skip to Content
Merck
All Photos(3)

Key Documents

MABN754

Sigma-Aldrich

Anti-PAH Antibody, clone 6H10.1

clone 6H10.1, from mouse

Synonym(s):

Phenylalanine-4-hydroxylase, PAH, Phe-4-monooxygenase

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€429.00

€429.00


Estimated to ship on16 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€429.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€429.00


Estimated to ship on16 April 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

6H10.1, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PAH(5053)

General description

PAH, also known as Phenylalanine-4-hydroxylase , Phe-4-monooxygenase, and encoded by the gene name PAH, belongs to the biopterin-dependent aromatic amino acid hydroxylase family. Phenylalanine hydroxylase is the rate-limiting enzyme of the metabolic pathway that degrades excess phenylalanine. Phenylalanine hydroxylase (PheOH, alternatively PheH or PAH) is an enzyme that catalyzes the hydroxylation of the aromatic side-chain of phenylalanine to generate tyrosine. PheOH is one of three members of the pterin-dependent amino acid hydroxylases, a class of monooxygenase that uses tetrahydrobiopterin (BH4, a pteridine cofactor) and a non-heme iron for catalysis. During the reaction, molecular oxygen is heterolytically cleaved with sequential incorporation of one oxygen atom into BH4 and phenylalanine substrate. PAH has been associated with Phenylketonuria PKU, an autosomal recessive inborn error of phenylalanine metabolism, due to severe phenylalanine hydroxylase deficiency. Additioanlly, PAH has been associated with Non-phenylketonuria hyperphenylalaninemia (Non-PKU HPA), a mild form of phenylalanine hydroxylase deficiency characterized by phenylalanine levels persistently below 600 mumol, which allows normal intellectual and behavioral development without treatment. Finally, PAH may play a role in the Hyperphenylalaninemia (HPA), a mildest form of phenylalanine hydroxylase deficiency. PAH is broadly expressed, with greatest levels in skeletal muscle followed by heart, brain, pancreas and testis.

Immunogen

GST-tagged recombinant protein corresponding to human PAH.

Application

This Anti-PAH antibody is validated for use in WB, IH for the detection of PAH.
Western Blotting Analysis: 1.0 µg/mL from a representative lot detected PAH in 10 µg of human liver tissue lysate.
Immunohistochemistry Analysis: A 1:50-250 dilution from a representative lot detected PAH in human cerebral cortex and human liver tissue.

Quality

Evaluated by Western Blotting in HepG2 cell lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected PAH in 10 µg of HepG2 cell lysate.

Target description

~52 kDa observed

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service