MABN718
Anti-SMAD5 Antibody, clone 3H9
ascites fluid, clone 3H9, from mouse
Synonym(s):
Mothers against decapentaplegic homolog 5, MAD homolog 5, Mothers against DPP homolog 5, SMAD family member 5, SMAD5
Select a Size
About This Item
biological source
mouse
Quality Level
antibody form
ascites fluid
antibody product type
primary antibodies
clone
3H9, monoclonal
species reactivity
rat, human
technique(s)
flow cytometry: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
western blot: suitable
isotype
IgG1
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... SMAD5(4090)
General description
Immunogen
Application
Immunohistochemistry Analysis: A 1:200-1,000 dilution from a representative lot detected SMAD5 in human brain and lung cancer tissues.
Flow Cytometry Analysis: A 1:200-400 dilution from a representative lot detected SMAD5 in Jurkat cells.
Optimal working dilutions must be determined by end user.
Neuroscience
Signaling Neuroscience
Quality
Western Blotting Analysis: A 1:500-2,000 dilution of this antibody detected SMAD5 in HeLa, SK-N-SH, PC-12, Jurkat, and K562 cell lysates.
Target description
Physical form
Storage and Stability
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Analysis Note
HeLa, SK-N-SH, PC-12, Jurkat, and K562 cell lysates
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service