Skip to Content
Merck
All Photos(2)

Key Documents

MABN671

Sigma-Aldrich

Anti-DLK1 Antibody, clone 3A10

ascites fluid, clone 3A10, from mouse

Synonym(s):

Protein delta homolog 1, DLK-1, FA1, ZOG, pG2, PREF1, Pref-1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€411.00

€411.00


Estimated to ship on22 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μL
€411.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€411.00


Estimated to ship on22 April 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

3A10, monoclonal

species reactivity

mouse, human

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable

isotype

IgG1

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... DLK1(8788)

General description

Protein delta homolog 1or DLK-1 (DLK1), also known as pG2 and the Preadipocyte factor-1 (Pref-1), is a single pass type 1 membrane protein known to play a steering role in neuroendocrine differentiation. The protein is expressed within the stromal cells in contact with the vascular structure of the placental villi, yok sac, fetal liver, adrenal cortex and pancreas during development and DLK1 is also expressed within the beta cells of the islets of Langerhans in the adult. DLK1 plays an important gene regulatory expression role during the development of not only these very early structures but also in the regulation of the development of many specialized cells and tissues such as adipocytes. In adipose tissue, DLK1/Pref-1 is specifically expressed in preadipocytes but not in adipocytes and thus can be used as a preadipocyte marker. The inhibition operates through the creation of a soluble cleaved cytoplasmic form of DLK1 via the action of the metalloproteinase ADAM17 on mature DLK1. Fetal Antigen 1 (FA1) or soluble DLK-1/Pref-1 activates the MAPK kinase/ERK pathway, and via Sox9 interactions ultimately inhibits the differentiation of pre-adipocytes into mature adipocytes. Furthermore, by inducing Sox9, Pref-1 promotes chondrogenic induction of mesenchymal cells but prevents chondrocyte maturation as well as osteoblast differentiation. Thus, DLK1/Pref-1 directs multipotent mesenchymal cells (MSCs) toward the chondrogenic lineage but inhibits differentiation into adipocytes as well as osteoblasts and chondrocytes. Naturally, DLK1 has multiple isoforms each with slightly differing but related influences on MSCs development. Finally, DLK1 is also expressed in various tumors including small cell lung carcinoma and numerous neuroendocrine cancers.

Immunogen

Purified recombinant fragment of human DLK1 expressed in E. Coli.

Application

Detect DLK1 using this mouse monoclonal antibody, Anti-DLK1 Antibody, clone 3A10 validated for use in IHC & Immunofluorescence.
Immunofluorescence Analysis: A 1:200-1,000 dilution from a representative lot detected DLK1 in U251 cells.

Optimal working dilutions must be determined by end user.

Quality

Evaluated by Immunohistochemistry Analysis in human pancreas tissue.

Immunohistochemistry Analysis: A 1:100 dilution of this antibody detected DLK1 in human pancreas tissue.

Target description

45 calculated

Analysis Note

Control
Human pancreas tissue

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Susmita Kaushik et al.
Science advances, 8(46), eabq2733-eabq2733 (2022-11-17)
Adipogenesis is a tightly orchestrated multistep process wherein preadipocytes differentiate into adipocytes. The most studied aspect of adipogenesis is its transcriptional regulation through timely expression and silencing of a vast number of genes. However, whether turnover of key regulatory proteins

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service