Skip to Content
Merck
All Photos(1)

Key Documents

MABN668

Sigma-Aldrich

Anti-Ly6C Antibody, clone 6C3

clone 6C3, from rat

Synonym(s):

Lymphocyte antigen 6C2, Ly-6C2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€377.00

€377.00


Estimated to ship on29 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μL
€377.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€377.00


Estimated to ship on29 April 2025Details


Request a Bulk Order

biological source

rat

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

6C3, monoclonal

species reactivity

mouse

technique(s)

dot blot: suitable
flow cytometry: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Ly6C1(17067)

General description

Lymphocyte antigen 6C2 (Ly6C) is a 131 AA long membrane protein that is linked to macrophage phenotype according to expression levels.

Immunogen

Whole cells expressing mouse Ly6C.

Application

Detect Lymphocyte antigen 6C2 using this rat monoclonal antibody, Anti-Ly6C Antibody, clone 6C3 validated for use in Flow Cytometry & Dot Blot.
Dot Blot Analysis: A representative lot from an independent laboratory detected Ly6C in bone marrow-derived monocytes expressing 6C3 (Butovsky, O., et al. (2012). J Clin Invest. 122(9):3063-3087.).
Research Category
Neuroscience
Research Sub Category
Signaling Neuroscience

Quality

Evaluated by Flow Cytometry in mouse spleen cells.

Flow Cytometry Analysis: 1 µg of this antibody detected Ly6C in 1X10E6 mouse spleen cells.

Target description

14 kDa calculated

Physical form

Format: Purified
Protein G Purified
Purified rat monoclonal IgG2aκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oleg Butovsky et al.
Nature neuroscience, 17(1), 131-143 (2013-12-10)
Microglia are myeloid cells of the CNS that participate both in normal CNS function and in disease. We investigated the molecular signature of microglia and identified 239 genes and 8 microRNAs that were uniquely or highly expressed in microglia versus
Zhilong Jiang et al.
Respiratory research, 18(1), 217-217 (2017-12-30)
SOCS3 (suppressor of cytokine signaling 3) is a negative regulator of JAK/STAT3 signaling pathway and participates in the regulation of lung inflammation in a mouse model with acute lung injury (ALI). However, it is not well understood how SOCS3 regulates
Qin Huang et al.
PloS one, 14(11), e0225206-e0225206 (2019-11-15)
The engineered AAV-PHP.B family of adeno-associated virus efficiently delivers genes throughout the mouse central nervous system. To guide their application across disease models, and to inspire the development of translational gene therapy vectors for targeting neurological diseases in humans, we

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service