Skip to Content
Merck
All Photos(1)

Key Documents

MABN643

Sigma-Aldrich

Anti-NDUFS3 Antibody, clone 6H7.1

clone 6H7.1, from mouse

Synonym(s):

NADH dehydrogenase [ubiquinone] iron-sulfur protein 3, mitochondrial, Complex I-30kD, CI-30kD, NADH-ubiquinone oxidoreductase 30 kDa subunit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€306.00

€306.00


Estimated to ship on11 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€306.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€306.00


Estimated to ship on11 April 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

6H7.1, monoclonal

species reactivity

rat, human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... NDUFS3(4722)

General description

NDUFS3 is the core subunit of the mitochondrial respiratory chain′s NADH dehydrogenase complex, also known as Complex I, the complex critical for the production of ATP and NADH electron transfer. In mammals Complex I consists of some 45 different proteins, NDUFS3 is the critical core subunit of the complex. Expressed in all cells and localized in the mitochondria but encoded by a nuclear gene, NDUFS3 plays a critical role in electron transport, our internal cellular respiratory chain, and mitochondrial function. NDUFS3 also is important in disease phenotypes and various cancers. Mutations in NDUFS3 give rise to Leigh syndrome a mitochondrial dysfunctional disease. In cancers, down regulation of NDUFS3 is linked with lowered host survival and increased cancer pathology.

Immunogen

GST-tagged recombinant protein corresponding to human NDUFS3.

Application

Immunohistochemistry Analysis: A 1:250 dilution from a representative lot detected NDUFS3 in human cerebral cortex, rat hippocampus, and rat cerebellum tissue.
Research Category
Neuroscience
Research Sub Category
Developmental Signaling
This Anti-NDUFS3 Antibody, clone 6H7.1 is validated for use in Western Blotting and Immunohistochemistry (Paraffin) for the detection of NDUFS3.

Quality

Evaluated by Western Blotting in THP-1 cell lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected NDUFS3 in 200 µg of THP-1 cell lysate.

Target description

~30 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service