Skip to Content
Merck
All Photos(1)

Documents

MABN467

Sigma-Aldrich

Anti-DLX-1 Antibody, clone L43/40

clone L43/40, from mouse

Synonym(s):

Homeobox protein DLX-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

L43/40, monoclonal

species reactivity

human

species reactivity (predicted by homology)

rat (based on 100% sequence homology), mouse (based on 100% sequence homology)

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... DLX1(1745)

General description

Homeobox protein DLX-1 (DLX-1) is a developmental potein that is localized within the cell nucleus. DLX-1 likely has regulatory roles in the development of the ventral forebrain, craniofacial patterning and morphogenesis, and may be involved in the early development of diencephalic subdivisions.

Immunogen

Linear peptide corresponding to human DLX-1.

Application

Detect Dlx 1 using this mouse monoclonal antibody, Anti-DLX-1 Antibody, clone L43/40 validated for use in western blotting & IHC.
Immunohistochemistry Analysis: A representative lot detected DLX-1 in rat hippocampus tissue (Prof. J. Trimmer, University of California, Davis.).

Western Blotting Analysis: A representative lot detected DLX-1 in whole cell extracts of wild type embryonic day 15 telencephalons and demonstrated a loss of signal in whole cell extracts of embryonic day 15 telencephalons from DLX1/2 knock out mice (Prof. J. Trimmer, University of California, Davis.).
Research Category
Neuroscience
Research Sub Category
Developmental Neuroscience

Quality

Evaluated by Western Blotting in HepG2 cell lysates.

Western Blotting Analysis: 1 µg/mL of this antibody detected DLX-1 in 10 µg of HepG2 cell lysate.

Target description

~27 kDa observed. Uncharacterized band(s) may be observed in some cell lysates.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2aκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HepG2 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service