Skip to Content
Merck
All Photos(1)

Key Documents

MABN449

Sigma-Aldrich

Anti-Protein capicua homolog (CIC), clone 6E12.1 Antibody

clone 6E12.1, 1 mg/mL, from mouse

Synonym(s):

Protein capicua homolog, Protein capicua homolog (CIC)

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€377.00

€377.00


Estimated to ship on22 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€377.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€377.00


Estimated to ship on22 April 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

6E12.1, monoclonal

species reactivity

human, rat

concentration

1 mg/mL

technique(s)

immunohistochemistry: suitable (paraffin)

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CIC(23152)

General description

Protein capicua homolog (CIC) , also known as Capicua Transcriptional Repressor, Capicua Homolog, and encoded by the gene CIC, is a transcriptional repressor which plays a role in development of the central nervous system (CNS). CIC is a new subfamily of the HMG-box superfamily and the gene′s expression is predominantly restricted to immature granule cells in the cerebellum, hippocampus and olfactory bulb in the CNS, and thus the gene is implicated in CNS development, in particular in granule cell development. CIC appears to be central to cerebellar development and ErbB signaling as well. Recent research has demonstrated that CIC is highly expressed in certain cancers including medulloblastomas, but also the loss of expression may serve as a biomarker for other cancer types such as oligodendrogliomas.

Immunogen

GST-tagged recombinant protein corresponding to human Protein capicua homolog (CIC).

Application

Immunohistochemistry Analysis: A 1:500-2,000 dilution from a representative lot detected Protein capicua homolog (CIC) in human cerebral cortex, rat olfactory bulb, and rat cerebral cortex tissue.
Research Category
Neuroscience
Research Sub Category
Developmental Signaling
This Anti-Protein capicua homolog (CIC), clone 6E12.1 Antibody is validated for use in Immunohistochemistry (Paraffin) for the detection of Protein capicua homolog (CIC).

Quality

Evaluated by Immunohistochemistry in rat cerebellum tissue.

Immunohistochemistry Analysis: A 1:500 dilution of this antibody detected Protein capicua homolog (CIC) in rat cerebellum tissue.

Target description

164 kDa calculated

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Severa Bunda et al.
Nature communications, 10(1), 661-661 (2019-02-10)
Capicua (CIC) is a transcriptional repressor that counteracts activation of genes downstream of receptor tyrosine kinase (RTK)/Ras/ERK signaling. It is well-established that tumorigenesis, especially in glioblastoma (GBM), is attributed to hyperactive RTK/Ras/ERK signaling. While CIC is mutated in other tumors
Severa Bunda et al.
Molecular cancer research : MCR, 18(5), 774-786 (2020-02-08)
Capicua (CIC) is a transcriptional repressor that counteracts activation of genes in response to receptor tyrosine kinase (RTK)/Ras/ERK signaling. Following activation of RTK, ERK enters the nucleus and serine-phosphorylates CIC, releasing it from its targets to permit gene expression. We

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service