Skip to Content
Merck
All Photos(2)

Key Documents

MABN358

Sigma-Aldrich

Anti-NB thymosin beta (TMSB15A Antibody), Clone 4A6.3

clone 4A6.3, from mouse

Synonym(s):

Thymosin beta-15A, NB thymosin beta, Thymosin-like protein 8

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€408.00

€408.00


Estimated to ship on17 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€408.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€408.00


Estimated to ship on17 April 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

4A6.3, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin)

isotype

IgMκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... TMSB15A(11013)

General description

TMSB15A protein is a key regulator of the cytoskeleton organization and polymerization, and it thus plays a role in cell migration, proliferation and cancer. TMSB15A does this because TMSB15A binds to and sequesters actin monomers and limits action polymerization. TMSB15A is differentially expressed in many tissues and cancer cell lines. Because of differential expression in certain cancers, for instance in triple negative breast cancer but not colon cancer, TMSB15A is being examined as a histochemical marker for measuring the effectiveness of cancer therapy on cancer. TMSB15A is also important in the nervous system and brain diseases. Notably, TMSB15A expression is increased upon brain stress and surprisingly TMSB15A localization differs depending upon the type of stress used and neuronal cell type. Because of its interaction with actin, TMSB15A is localized to the cytoplasm most of time, however, in the brain and when cells are placed under various stresses, TMSB15A can be translocated to the brain to perform yet unknown functions as well.

Immunogen

KLH-conjugated linear peptide corresponding to Human TMSB15A.

Application

Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected TMSB15A in human thalamic and human brain cancer tissues.
This Anti-NB thymosin beta (TMSB15A Antibody), Clone 4A6.3 is validated for use in Immunohistochemistry (Paraffin) for the detection of NB thymosin beta (TMSB15A).

Quality

Evaluated by Immunohistochemistry in human cerebral cortex tissue.

Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected TMSB15A in human cerebral cortex tissue

Target description

5 kDa calculated

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service