Skip to Content
Merck
All Photos(2)

Key Documents

MABN281

Sigma-Aldrich

Anti-Numb Antibody, clone 6G10.1

clone 6G10.1, from mouse

Synonym(s):

Protein numb homolog, h-Numb, S171

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€325.00

€325.00


Estimated to ship on07 March 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€325.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€325.00


Estimated to ship on07 March 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

6G10.1, monoclonal

species reactivity

human

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgMκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... NUMB(8650)

General description

Numb plays a key role in neurogenesis and in cell fate decisions. It maintains self-renewal properties in neural progenitor cells, which are also called radial glial cells (RGCs). It functions through inhibition of Notch signaling. Additionally, it is suggested to mediate local repair in brain ventricular wall damage.

Specificity

This antibody recognizes Numb isoforms 1 and 2.

Immunogen

KLH-conjugated linear peptide corresponding to human Numb isoforms 1 and 2.

Application

Detect Numb using this Anti-Numb Antibody, clone 6G10.1 validated for use in WB & IC.
Immunocytochemistry Analysis: A 1:500 dilution from a representative lot detected Numb in HeLa and A431 cells.
Research Category
Neuroscience
Research Sub Category
Developmental Neuroscience

Quality

Evaluated by Western Blot in A431 cell lysate.

Western Blot Analysis: 1 µg/mL of this antibody detected Numb in 10 µg of A431 cell lysate.

Target description

~70 kDa observed. Uniprot describes four isoforms at ~71 kDa (isoform 1), ~66 kDa (isoform 2), ~69 kDa (isoform 3), and ~65 kDa (isoform 4).

Physical form

Format: Purified
Purified mouse monoclonal IgMκ in buffer containing PBS with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
A431 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service