Skip to Content
Merck
All Photos(1)

Key Documents

MABN2655

Sigma-Aldrich

Anti-Kynurenine Antibody, clone 11F9

Synonym(s):

(2S)-2-Amino-4-(2-aminophenyl)-4-oxo-butanoic acid

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
€137.00
100 μG
€347.00

€137.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 μG
€137.00
100 μG
€347.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€137.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

11F9, monoclonal

mol wt

calculated mol wt 208.22 kDa
observed mol wt ~65 kDa

purified by

using protein G

species reactivity

human

species reactivity (predicted by homology)

all

packaging

antibody small pack of 100 μg

technique(s)

western blot: suitable

isotype

IgG1κ

epitope sequence

Unknown

Protein ID accession no.

UniProt accession no.

storage temp.

2-8°C

General description

Kynurenine, a breakdown product of tryptophan, is produced by the action of tryptophan dioxygenase mainly in the liver. Kynurenine and its breakdown products perform several biological functions, including dilating blood vessels during inflammation and regulating the immune response. The kynurenine pathway is reported to be involved in the synthesis of NAD+ from the L-tryptophan and alterations in this pathway have been linked to several neurological and psychiatric disorders. Kynurenine and its metabolites can readily pass-through cell membranes and can diffuse through the cortex of the lens. Kynurenine plays a protective role in the eye by shielding the retina from UV damage. However, it can also cause damage to lens proteins if glutathione levels are not sufficient to prevent the reaction of kynurenines with proteins. Kynurenines undergo spontaneous deamination to form ,β-unsaturated ketones that are highly reactive with nucleophilic amino acids in lens proteins. Kynurenine levels decrease with age and with cataract formation, which is mainly via its reaction with lens proteins. Clone 11F9 detects age-related changes in lens proteins altered by kynurenines. It specifically recognizes kynurenine modified water soluble human lens proteins, but not the ones with other tryptophan oxidation products. (Ref.: Mailankot, M, and Nagaraj, RH. (2010). Int. J. Biochem. Cell Biol. 42(9); 1446-1454; Staniszewska, MM., and Nagaraj, RH. (2007). J. Immunol. Methods. 324; 63-73; Staniszewska, MM., and Nagaraj, RH. (2005). J. Biol. Chem. 280(23); 22154-22164).

Specificity

Clone 11F9 is a mouse monoclonal antibody that detects kynurenine tagged proteins.

Immunogen

BSA-conjugated N-formylkynurenine

Application

Quality Control Applications

Evaluated by Western Blotting with Kynurenine modified Bovine serum albumin (BSA).

Western Blotting Analysis (WB): A 1:20,000 dilution of this antibody detected Kynurenine modified BSA, but not the unmodified BSA.

Tested applications

ELISA Analysis: A representative lot detected Kynurenine in ELISA applications (Staniszewska, M., et al. (2007). J Immunol Methods. 324(1-2); 63-73).

Immunocytochemistry Analysis: A representative lot detected Kynurenine in Immunocytochemistry applications (Staniszewska, M., et al. (2007). J Immunol Methods. 324(1-2); 63-73).

Western Blotting Analysis: A representative lot detected Kynurenine in Western Blotting applications (Staniszewska, M., et al. (2007). J Immunol Methods. 324(1-2); 63-73).

Immunohistochemistry Applications: A representative lot detected Kynurenine in Immunohistochemistry applications (Staniszewska, M., et al. (2007). J Immunol Methods. 324(1-2); 63-73).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Physical form

Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Recommended storage: +2°C to +8°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service