Skip to Content
Merck
All Photos(4)

Key Documents

MABN2639

Sigma-Aldrich

Anti-AIDA1 Antibody, clone 1A11

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
€145.00
100 μG
€366.00

€145.00


Estimated to ship on19 February 2025Details


Request a Bulk Order

Select a Size

Change View
25 μG
€145.00
100 μG
€366.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€145.00


Estimated to ship on19 February 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1A11, monoclonal

mol wt

calculated mol wt 139.21, 48.37, and 57.81 kDa
observed mol wt ~160, 57, and 50 kDa

purified by

using protein G

species reactivity

rat

species reactivity (predicted by homology)

monkey, canine, feline, human, bovine

packaging

antibody small pack of 100 μg

isotype

IgG2bκ

epitope sequence

C-terminal half

UniProt accession no.

shipped in

2-8°C

target post-translational modification

unmodified

General description

Ankyrin repeat and sterile alpha motif domain-containing protein 1B (UniProt: P0C6S7; also known as Amyloid-beta protein intracellular domain-associated protein 1, AIDA-1, E2A-PBX1-associated protein, EB-1) is encoded by the Anks1b gene (Gene ID: 314721) in rat. AIDA-1 is an adaptor protein that has been implicated in Cajal body and nucleolar function. It is expressed throughout the brain and is highly expressed in the hippocampus. It is one of the most abundant proteins at postsynaptic densities where it is reported to bind to PSD95 and associates with NMDA receptors (NMDARs). In the brain it interacts with amyloid b precursor protein and plays a role in brain development. AIDA-1 preferentially associates with GluN2B-containing NMDARs and plays a role in their transport from the endoplasmic reticulum (ER). Loss of AIDA-1 results in the accumulation of GluN2B in the ER and decreases GluN2B and increases GluN2A distribution at the synapse. However, loss of AIDA-1 does not affect AMPAR-mediated synaptic transmission or the probability of glutamate release. Mutations in Anks1b gene are known to affect GluN2B function and are linked to the pathogenesis of neuropsychiatric disorders. (Ref.: Carbonell, AU., et al., (2019). Nat. Commun. 10(1); 3529; Tindi, JO., et al. (2015). J. Neurosci. 35(24); 8986- 8996).

Specificity

Clone 1A11 is a mouse monoclonal antibody that detects AIDA-1 in multiple mammalian species. It targets an epitope within 10 amino acids from the SAM2 domain in the C-terminal half.

Immunogen

A linear peptide corresponding to 10 amino acids from the SAM2 domain within the C-terminal half of rat Ankyrin repeat and sterile alpha motif domain-containing protein 1B (AIDA-1).

Application

Quality Control Testing

Evaluated by Western Blotting in Rat brain tissue lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected AIDA1 in Rat brain tissue lysate.

Tested Applications

Western Blotting Analysis: A representative lot detected AIDA1 in Western Blotting applications (Tindi, J.O., et al. (2015). J Neurosci. 35(24): 8986-96).

Immunofluorescence Analysis: A representative lot detected AIDA1 in Immunofluorescence applications (Tindi, J.O., et al. (2015). J Neurosci. 35(24): 8986-96).

Immunohistochemistry (Paraffin) Analysis (IHC(P)): A 1:50 dilution from a representative lot detected AIDA1 in rat cerebral cortex, hippocampus, and cerebellum tissue sections.

Immunoprecipitation Analysis: A representative lot immunoprecipitated AIDA1 in Immunoprecipitation applications (Carbonell, A.U., et al. (2019). Nat Commun. 10(1): 3529).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-AIDA1, clone 1A11, Cat. No. MABN2639, is a mouse monoclonal antibody that detects AIDA1/ANKS1B and is tested for use in Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, and Western Blotting.

Physical form

Purified mouse monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Recommend storage at +2°C to +8°C. For long term storage antibodies can be kept at -20°C. Avoid repeated freeze-thaws.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service