Skip to Content
Merck
All Photos(1)

Key Documents

MABN2430M

Sigma-Aldrich

Anti-Kallikrein 7 (KLK7) Antibody, clone 83-1

clone 83-1, from mouse

Synonym(s):

Serine protease 6, EC 3.4.21.117, hK7, Stratum corneum chymotryptic enzyme, hSCCE

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€317.00

€317.00


Estimated to ship on05 May 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€317.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

€317.00


Estimated to ship on05 May 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

83-1, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μg

technique(s)

ELISA: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... KLK7(5650)

General description

Kallikrein-7 (UniProt: P49862; also known as hK7, KLK7, Serine protease 6, Stratum corneum chymotryptic enzyme, hSCCE) is encoded by the KLK7 (also known as PRSS6, SCCE) gene (Gene ID: 5650) in human. KLK7 is a chymotrypsin-like serine protease of the member of peptidase S1 family. It is synthesized with a signal peptide of 22 amino acids (aa 1-22) and a propeptide region (aa 23-29) that are subsequently cleaved to generate the mature form. Human KLK7 displays about 40 to 60% amino acid homology with other members of the family. It cleaves proteins with aromatic side chains in the P1 position. KLK7 is abundantly expressed in the skin and is also present in the brain, mammary gland, cerebellum, spinal cord, and kidney. Lower levels of expression have been reported in in salivary glands, uterus, thymus, thyroid, placenta, trachea, and testis. Its levels are shown to be up-regulated in ovarian carcinoma, especially late-stage serous carcinoma. Less differentiated tumors of more advanced stage show higher level KLK7 expression. KLK7 activity is inhibited by zinc and copper ions.

Specificity

Clone 83-1 detects Kallikrein -7 in human tissues.

Immunogen

Full-length human recombinant Kallikrein-7.

Application

Anti-Kallikrein 7 (KLK7), clone 83-1, Cat. No. MABN2430, is a highly specific mouse monoclonal antibody that targets Kallikrein-7 and has been tested in ELISA and Western Blotting.
ELISA Analysis: A representative lot detected Kallikrein 7 (KLK7) in ELISA applications (Shan, S.J., et. al. (2006). Clin Chem. 52(10):1879-86; Kishi, T., et. al. (2004). Clin Chem. 50(4):709-16).
Research Category
Neuroscience

Quality

Evaluated by Western Blotting in human brain tissue lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected Kallikrein 7 (KLK7) in 10 µg of human brain tissue lysate.

Target description

~27 kDa observed; 27.53 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Sodium bicarbonate buffer, pH 8.2 with 0.02% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service