Skip to Content
Merck
All Photos(1)

Key Documents

MABN2418

Sigma-Aldrich

Anti-CELF4 Antibody, clone N446/80

clone N446/80, from mouse

Synonym(s):

CUGBP Elav-like family member 4, Bruno-like protein 4, CUG-BP- and ETR-3-like factor 4, RNA-binding protein BRUNOL-4

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
€130.00
100 μL
€350.00

€130.00


Estimated to ship on08 April 2025Details


Request a Bulk Order

Select a Size

Change View
25 μL
€130.00
100 μL
€350.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€130.00


Estimated to ship on08 April 2025Details


Request a Bulk Order

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

N446/80, monoclonal

species reactivity

human, rat, mouse

packaging

antibody small pack of 25 μL

technique(s)

western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... CELF4(56853)

General description

CUGBP Elav-like family member 4 (UniProt: Q9BZC1; also known as CELF-4, Bruno-like protein 4, CUG-BP- and ETR-3-like factor 4, RNA-binding protein BRUNOL-4) is encoded by the CELF4 (also known as BRUNOL4) gene (Gene ID: 56853) in human. CELF-4 is a RNA-binding protein that is implicated in the regulation of pre-mRNA alternative splicing. It is ubiquitous in its distribution, but is strongly expressed in the cerebellum, hippocampus, amygdala, temporal and frontal cortex, and frontal lobes. It mediates exon inclusion and/or exclusion in pre-mRNA that are subject to tissue-specific and developmentally regulated alternative splicing. It is shown to specifically activate exon 5 inclusion of cardiac isoforms of TNNT2 during heart remodeling at the juvenile to adult transition. CELF-4 is reported to promote exclusion of both the smooth muscle and non-muscle exons in actinin pre-mRNAs. It binds to muscle-specific splicing enhancer (MSE) intronic sites flanking the alternative exon 5 of TNNT2 pre-mRNA. The N-terminal region (aa 1-298) is considered to be sufficient for its RNA-binding and MSE-dependent splicing activity. CELF-4 contains contain two N-terminal RNA recognition motif (RRM) domains (aa 54-135 and 152-232) and one C-terminal RRM domain (aa 404-479). Five isoforms of CELF-4 have been described that are produced by alternative splicing.

Specificity

Clone N446/80 specifically detects CUGBP Elav-like family member 4 (CELF4). It targets an epitope with in 86 amino acids from the N-terminal region. It does not react with CELF6.

Immunogen

A recombinant fragment corresponding to 86 amino acids from the N-terminal region of human CUGBP Elav-like family member 4 (CELF-4).
Epitope: N-terminus

Application

Anti-CELF4, clone N446/80, Cat. No. MABN2418, is a highly specific mouse monoclonal antibody that targets CUGBP Elav-like family member 4 and has been tested for use in Western Blotting.
Research Category
Neuroscience

Quality

Evaluated by Western Blotting in human hippocampus tissue lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected CELF4 in human hippocampus tissue lysate.

Target description

~46 kDa observed; 51.97 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service