Skip to Content
Merck
All Photos(1)

Key Documents

MABN2415

Sigma-Aldrich

Anti-phospho-NHE3 (Ser552) Antibody, clone 14D5

clone 14D5, from mouse

Synonym(s):

Sodium/hydrogen exchanger 3, Na(+)/H(+) exchanger 3, Solute carrier family 9 member 3, NHE-3

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€373.00

€373.00


Estimated to ship on04 June 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€373.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€373.00


Estimated to ship on04 June 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

14D5, monoclonal

species reactivity

rat, opossum, bovine, mouse, human

technique(s)

ELISA: suitable
immunofluorescence: suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

phosphorylation (pSer552)

Gene Information

General description

Sodium/hydrogen exchanger 3 (UniProt: P26433; also known as Na(+)/H(+) exchanger 3, NHE-3, Solute carrier family 9 member 3) is encoded by the Slc9a3 (Nhe3) gene (Gene ID: 24784) in rat. NHE-3 is a multi-pass membrane protein that is involved in pH regulation to eliminate acids generated by active metabolism or to counter adverse environmental conditions. It is a major proton extruding system driven by the inward sodium ion chemical gradient. NHE-3 is shown to be most abundant in colon and small intestine and is also present in stomach and in proximal tubes and outer medulla of kidney. In kidneys it mediates the majority of sodium bicarbonate reabsorption by the proximal tubules. NHE-3 can be phosphorylated and regulated by multiple kinases, including PKA, SGK1, and CK2 and its phosphorylation is critical in its regulation. Parathyroid hormone is shown to enhance PKA-mediated phosphorylation of Serine 552, however, this phosphorylation does not affect the activity of NHE-3. (Ref.: Kocinsky, HS., et al. (2007). Am J Physiol Renal Physiol 293 (1); F212-F218).

Specificity

This mouse monoclonal antibody (clone 14D5) specifically detects Sodium/hydrogen exchanger 3 phosphorylated on Serine 552.

Immunogen

KLH-conjugated linear peptide corresponding to 22 amino acids surrounding phospho Serine 552 from the C-terminal half of rat Sodium/hydrogen exchanger 3 (NHE3).

Application

Anti-phospho-NHE3 (Ser552), clone 14D5, Cat. No. MABN2415, is a highly specific mouse monoclonal antibody that targets Sodium/hydrogen exchanger 3 and has been tested for use in ELISA, Immunofluorescence, and Western Blotting.
Immunofluorescence Analysis: A representative lot detected phospho-NHE3 (Ser552) in Immunofluorescence applications (Kocinsky, H.S., et. al. (2005). Am J Physiol Renal Physiol. 289(2):F249-58).

Western Blotting Analysis: A representative lot detected phospho-NHE3 (Ser552) in Western Blotting applications (Kocinsky, H.S., et. al. (2005). Am J Physiol Renal Physiol. 289(2):F249-58; Kocinsky, H.S., et. al. (2007). Am J Physiol Renal Physiol. 293(1):F212-8).

ELISA Analysis: A representative lot detected phospho-NHE3 (Ser552) in ELISA applications (Kocinsky, H.S., et. al. (2005). Am J Physiol Renal Physiol. 289(2):F249-58).
Research Category
Neuroscience

Quality

Evaluated by Western Blotting in COS-7 cells either transfected with wild type NHE3, mutated NHE3 (S552A), or untransfected COS-7 (WT) cell lysates.

Western Blotting Analysis: 2 µg/mL of this antibody detected Sodium/hydrogen exchanger 3 in lysate from COS-7 cells transfected with wild- type NHE-3 (pSer552). It did not detect NHE-3 in lysate from untransfected COS-7 cells or those transfected with mutated NHE-3 (S552A).


Target description

~85 kDa observed; 93.11 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yiran Han et al.
Frontiers in physiology, 13, 867244-867244 (2022-04-22)
Glycemic control is the key to the management of type 2 diabetes. Metformin is an effective, widely used drug for controlling plasma glucose levels in diabetes, but it is often the culprit of gastrointestinal adverse effects such as abdominal pain
Kayte A Jenkin et al.
Cellular and molecular gastroenterology and hepatology, 13(3), 695-716 (2021-11-26)
Diarrhea is one of the most common illnesses and is often caused by bacterial infection. Recently, we have shown that human Na+/H+ exchanger NHE3 (hNHE3), but not non-human NHE3s, interacts with the E3 ubiquitin ligase Nedd4-2. We hypothesize that this
Bruna Bezerra Lins et al.
Frontiers in physiology, 12, 642752-642752 (2021-04-20)
High plasma angiotensin II (Ang II) levels are related to many diseases, including hypertension, and chronic kidney diseases (CKDs). Here, we investigated the relationship among prolonged Ang II infusion/AT1 receptor (AT1R) activation, oxidative stress, and endoplasmic reticulum (ER) stress in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service