Skip to Content
Merck
All Photos(2)

Key Documents

MABN2406

Sigma-Aldrich

ANTI-TAU (MAPT) Antibody

mouse monoclonal, TNT-2

Synonym(s):

EC:4.6.1.2, Guanylate cyclase 2D, retinal, Guanylate cyclase E (GC-E), Microtubule-associated protein tau, Neurofibrillary tangle protein, Paired helical filament-tau, PHF-tau, RETGC-1, ROS-GC, Retinal guanylyl cyclase 1, Rod outer segment membrane guanylate cyclase

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€321.00

€321.00


Estimated to ship on10 June 2025Details

A recombinant, preservative-free antibody is available for your target. Try ZMS3420

Request a Bulk Order

Select a Size

Change View
100 μG
€321.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€321.00


Estimated to ship on10 June 2025Details

A recombinant, preservative-free antibody is available for your target. Try ZMS3420

Request a Bulk Order

Product Name

Anti-Tau Antibody, clone TNT-2, clone TNT-2, from mouse

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

TNT-2, monoclonal

mol wt

calculated mol wt 120.37 kDa
observed mol wt ~N/A kDa

purified by

using protein G

species reactivity

rat, human, mouse

packaging

antibody small pack of 100

technique(s)

ELISA: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG1κ

epitope sequence

C-terminal

NCBI accession no.

UniProt accession no.

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... MAPT(4137)
mouse ... Mapt(17762)
rat ... Mapt(29477)

General description

Microtubule-associated protein tau (UniProt: P10636; also known as Neurofibrillary tangle protein, Paired helical filament-tau, PHF-tau) is encoded by the MAPT (MAPTL, MTBT1, TAU) gene (Gene ID: 4137) in human. Tau proteins are mostly found in the axons of neurons, in the cytosol and in association with plasma membrane components. Normally, Tau promotes microtubule assembly and stability, and might be involved in the establishment and maintenance of neuronal polarity. The C-terminus binds axonal microtubules while the N-terminus binds neural plasma membrane components, suggesting that tau functions as a linker protein between both. Axonal polarity is predetermined by TAU/MAPT localization (in the neuronal cell) in the domain of the cell body defined by the centrosome. The localization of Tau protein in the Alzheimer s disease (AD) brain is markedly abnormal and may contribute to neuronal dysfunction. In AD, normal soluble cytoskeletal elements, such as Tau and neurofilaments, are transformed into insoluble paired helical filaments (PHFs). This is linked to the post-translational change in Tau, primarily its hyperphosphorylation by a number of protein kinases. In its non-phosphorylated state, Tau promotes rapid and extensive microtubule (MT) polymerization. A significant consequence of Tau hyperphosphorylation in AD is a reduction in its ability to bind microtubules and promote microtubule assembly. Hyperphosphorylated Tau may contribute to a destabilized microtubule network, impaired axonal transport, and ultimately result in NFT formation and neuronal death.

Specificity

Clone TNT-2 specirfically detects tau in human, mouse, and rat brain. It targets a sequence of 17 amino acids from the N-terminal region that is common to all isoforms (1-6) of tau.

Immunogen

KLH-conjugated linear peptide corresponding to 17 amino acids from the N-terminal region of human Tau.

Application

Quality Control Testing

Evaluated by Immunohistochemistry in mouse retina tissue section

Immunohistochemistry (Paraffin) Analysis (IHC): A 1:50 dilution from a representative lot detected Guanylate cyclase 1 in mouse retina tissue sections.

Tested Applications

Immunofluorescence Analysis: A representative lot detected Guanylate cyclase 1 in Immunofluorescence application (Ying, G., et al. (2018). J Biol Chem. 293(45):17546-17558).

Immunohistochemistry Analysis: A representative lot detected Guanylate cyclase 1 in Immunohistochemistry application (Haire, S.E., et al. (2006). Invest Ophthalmol Vis Sci. 47(9):3745-53).

Western Blotting Analysis: A representative lot detected Guanylate cyclase 1 in Western Blotting application (Haire, S.E., et al. (2006). Invest Ophthalmol Vis Sci. 47(9):3745-53).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Quality

Evaluated by Western Blotting in rat entorhinal cortex.

Western Blotting Analysis: 0.02 µg/mL of this antibody detected Tau in 10 µg of rat entorhinal cortex lysate.

Target description

~55 kda Observed. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Reconstitution

1.0 mg/mL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service