Skip to Content
Merck
All Photos(1)

Key Documents

MABN2395

Sigma-Aldrich

Anti-Peripherin-2 Antibody, clone 2B7

clone 2B7, from mouse

Synonym(s):

Retinal degeneration slow protein

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€388.00

€388.00


Estimated to ship on04 June 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€388.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€388.00


Estimated to ship on04 June 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2B7, monoclonal

species reactivity

mouse, bovine

technique(s)

immunocytochemistry: suitable
immunofluorescence: suitable
immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

mouse ... Prph2(19133)

General description

Peripherin-2 (UniProt: P15499; also known as Retinal degeneration slow protein) is encoded by the Prph2 (also known as Rds) gene (Gene ID: 19133) in murine species. Peripherin-2 is a multi-pass membrane glycoprotein found in the outer segment of both rod and cone photoreceptor cells. It is localized to the photoreceptor disc rim region and is required for the initial formation and subsequent maintenance of both rod and cone outer segments. It can function as an adhesion molecule for stabilization and compaction of outer segment disks and the maintenance of the curvature of the rim. Mutations Prph2 gene are reported to cause incurable retinal degeneration. Peripherin-2 can form a wide variety of covalent and non-covalent homo- and heteromeric complexes and failure to oligomerize properly can result in protein degradation and retinitis pigmentosa.

Specificity

Clone 2B7 detects peripherin-2 in murine and bovine eye. It targets an epitope within 23 amino acids from the C-terminal end.

Immunogen

KLH-conjugated linear peptide corresponding to 23 amino acids from the C-terminal region of murine Peripherin-2.

Application

Anti-Peripherin-2, clone 2B7, Cat. No. MABN2395, is a highly specific mouse monoclonal antibody that targets Peripherin-2/RDS and has been tested in Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), and Western Blotting.
Western Blotting Analysis: A representative lot detected Peripherin-2 in Western Blotting applications (Stuck, M.W., et. al. (2014). Hum Mol Genet. 23(23):6260-74).

Immunofluorescence Analysis: A representative lot detected Peripherin-2 in Immunofluorescence applications (Chakraborty, D., et. al. (2010). Hum Mol Genet. 19(24):4799-812).

Western Blotting Analysis: A representative lot detected Peripherin-2 in Western Blotting applications (Conley, S.M., et. al. (2010). Biochemistry. 49(5):905-11).

Immunocytochemistry Analysis: A representative lot detected Peripherin-2 in Immunocytochemistry applications (Conley, S.M., et. al. (2010). Biochemistry. 49(5):905-11).

Quality

Evaluated by Immunohistochemistry in the outer layer of photoreceptor cells of mouse eye tissue.

Immunohistochemistry Analysis: A 1:1,000 dilution of this antibody detected Peripherin-2 in the outer layer of photoreceptor cells of mouse eye tissue.

Target description

39.26 kDa calculated.

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ayse M Genc et al.
International journal of molecular sciences, 21(21) (2020-11-04)
The large number of inherited retinal disease genes (IRD), including the photopigment rhodopsin and the photoreceptor outer segment (OS) structural component peripherin 2 (PRPH2), has prompted interest in identifying common cellular mechanisms involved in degeneration. Although metabolic dysregulation has been

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service