Skip to Content
Merck
All Photos(1)

Key Documents

MABN1851

Sigma-Aldrich

Anti-Nav1.4 Na+ channel Antibody, clone N255_38

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
€122.00
100 μG
€308.00

€122.00


Estimated to ship on12 March 2025Details


Request a Bulk Order

Select a Size

Change View
25 μG
€122.00
100 μG
€308.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€122.00


Estimated to ship on12 March 2025Details


Request a Bulk Order

antibody form

purified antibody

clone

N255/38
monoclonal

purified by

using protein G

species reactivity

rat, mouse

concentration

(Please refer to lot specific datasheet.)

technique(s)

immunohistochemistry: suitable

isotype

IgG2aκ

UniProt accession no.

General description

Recombinant fragment corresponding to 118 amino acids from the N-terminal half of Rat Voltage-gated sodium channel subunit alpha (Nav1.4).

Specificity

Clone N255/38 is a mouse monoclonal antibody that targets Sodium channel protein type 4 subunit alpha (Nav1.4). It targets an epitope within the N-terminal, extracellular domain.

Application

Immunohistochemistry (Paraffin) Analysis: A 1:50 dilution from a representative lot detected Nav1.4 Na+ channel in rat cerebral cortex tissue sections.

Quality

Evaluated by Immunohistochemistry (Paraffin) in rat hippocampus tissue sections.

Immunohistochemistry (Paraffin) Analysis: A 1:50 dilution of this antibody detected Nav1.4 Na+ channel in rat hippocampus tissue sections.

Physical properties

208.8 kDa calculated.

Physical form

Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Rat.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service