Skip to Content
Merck
All Photos(1)

Key Documents

MABN1559

Sigma-Aldrich

Anti-ATBF1 Antibody, clone 5A1.1

clone 5A1.1, from mouse

Synonym(s):

Zinc finger homeobox protein 3, AT motif-binding factor 1, AT-binding transcription factor 1, Alpha-fetoprotein enhancer-binding protein, Zinc finger homeodomain protein 3, ZFH-3

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€330.00

€330.00


Estimated to ship on04 June 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€330.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€330.00


Estimated to ship on04 June 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

5A1.1, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μg

technique(s)

western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... ZFHX3(463)

General description

Zinc finger homeobox protein 3 (UniProt: Q15911; also known as AT motif-binding factor 1, AT-binding transcription factor 1, ATBF1, Alpha-fetoprotein enhancer-binding protein, Zinc finger homeodomain protein 3, ZFH-3) is encoded by the ZFHX3 (also known as ATBF1) gene (Gene ID: 463) in human. ATBF1 contains 23 C2H2-type zinc fingers and 4 homeobox DNA-binding domains. It is shown to translocate from the cytoplasm to the nucleus following TGF-beta stimulation. It acts as a transcriptional regulator that can act as an activator or a repressor. It inhibits the enhancer element of the AFP gene by binding to its AT-rich core sequence. In concert with SMAD-dependent TGF-beta signaling, it can repress the transcription of AFP via its interaction with SMAD2/3. ATBF1 is shown to regulate myoblasts differentiation through the binding to the AT-rich sequence of MYF6 promoter and promoter repression. It is not found in normal gastric mucosa, but is found in gastric carcinoma cells and its expression is higher in ER-positive breast tumors than ER-negative breast tumors. It is shown to down-regulate the MUC5AC promoter in gastric cancer cells.

Specificity

Clone 5A1.1 recognizes Zinc finger homeobox protein 3 (ATBF1) in human cells. It targets an epitope with in 87 amino acids from the C-terminal region. This antibody recognizes isoform of 306 and 404 kDa.

Immunogen

GST/His-tagged recombinant fragment corresponding to 87 amino acids from the C-terminal region of human Zinc finger homeobox protein 3.

Application

Anti-ATBF1, clone 5A1.1, Cat. No. MABN1559, is a highly specific mouse monoclonal antibody that targets Zinc finger homeobox protein 3 and has been tested in Western Blotting.

Quality

Evaluated by Western Blotting in SHSY-5Y cell lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected ATBF1 in 10 µg of SHSY-5Y cell lysate.


Target description

~404 kDa observed; 404.42 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service