Skip to Content
Merck
All Photos(2)

Key Documents

MABN124

Sigma-Aldrich

Anti-ATF-3 Antibody, clone 13A5.1

clone 13A5.1, from human(Recombinant)

Synonym(s):

Cyclic AMP-dependent transcription factor ATF-3, Activating transcription factor 3, cAMP-dependent transcription factor ATF-3

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€401.00

€401.00


Estimated to ship on22 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μL
€401.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€401.00


Estimated to ship on22 April 2025Details


Request a Bulk Order

biological source

human (Recombinant)

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

13A5.1, monoclonal

species reactivity

mouse, human

technique(s)

western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ATF3(467)

General description

Cyclic AMP-dependent transcription factor ATF-3 (UniProt P18847; also known as Activating transcription factor 3, cAMP-dependent transcription factor ATF-3) is encoded by the ATF3 gene (Gene ID 467) in human. ATF-3 belongs to the ATF/cAMP-responsive element-binding protein family of basic leucine zipper-type transcription factors. ATF-3 is markedly induced and functions as either a transcriptional activator or repressor in response to LPS, cytokines, hormones, DNA damage, and ER stress in various cell types. ATF3 plays key roles in atherosclerosis and hyperlipidemia, and is reported to negatively regulate the differentiation of adipocyte, chondrocyte, and osteoblast. ATF-3 dysfunction is closely related to various inflammatory diseases and cancer.

Specificity

Expected to react with spliced isoforms 1 and 5 (UniProt P18847-1 and P18847-5). Reactivity toward spliced isoforms 2, 3, and 4 (UniProt P18847-2 through P18847-4) of human ATF-3 has not been determined.

Immunogen

GST-tagged recombinant human ATF-3 fragment.

Application

Research Category
Neuroscience
Research Sub Category
Neurodegenerative Diseases
Western Blotting Analysis: A 1:1,000 dilution of this antibody detected ATF-3 in 10 µg of RAW264.7 cell lysate.

Quality

Evaluated by Western Blotting in THP-1 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected ATF-3 in 10 µg of THP-1 cell lysate.

Target description

~19 kDa observed.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2bκ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service