Skip to Content
Merck
All Photos(1)

Key Documents

MABN1100

Sigma-Aldrich

Anti-REG1A, clone 1A4 Antibody

ascites fluid, clone 1A4, from rabbit

Synonym(s):

Lithostathine-1-alpha, Islet cells regeneration factor, ICRF, Islet of Langerhans regenerating protein, REG, Pancreatic stone protein, PSP, Pancreatic thread protein, PTP, Regenerating islet-derived protein 1-alpha, REG-1-alpha, Regenerating protein I al

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

ascites fluid

antibody product type

primary antibodies

clone

1A4, monoclonal

species reactivity

human

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable

isotype

IgG1

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... REG1A(5967)

General description

Lithostathine-1-alpha, also known as Islet cells regeneration factor (ICRF) or Islet of Langerhans regenerating protein (REG) or Pancreatic stone protein (PSP) or Pancreatic thread protein (PTP) or Regenerating islet-derived protein 1-alpha (REG-1-alpha), and encoded by the gene name REG1A or PSPS or PSPS1 or REG is a member of the Reg family of secreted proteins with a C-type lectin domain. REG1A is associated with islet cell regeneration and diabetogenesis and may be involved in pancreatic lithogenesis. Due to variable glycosylation, pancreatic REG1A exists as multiple species of 16 - 18 kDa. REG1A promotes the maintenance and growth of pancreatic islet β-cells and intestinal villi and is upregulated in pancreatitis and some carcinomas. REG1A is an antigenic target in autoimmune diabetes.

Immunogen

Purified recombinant fragment of human REG1A fused with HlgGFc-tag expressed in HEK293 cells.

Application

Detect REG1A using this Anti-REG1A, clone 1A4 Antibody validated for use in IHC & immunofluorescence.
Immunofluorescence Analysis: A 1:200-1,000 dilution from a representative lot detected REG1A in PC12 cells.
Research Category
Neuroscience
Research Sub Category
Neurodegenerative Diseases

Quality

Immunohistochemistry Analysis: A 1:200-1,000 dilution from a representative lot detected REG1A in human pancreas tissue.

Target description

19 kDa calculated

Physical form

Mouse monoclonal IgG1 ascitic fluid containing up to 0.1% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service