Skip to Content
Merck
All Photos(2)

Key Documents

MABN109

Sigma-Aldrich

Anti-Synaptotagmin-4 Antibody, clone 10C11.1

clone 10C11.1, from mouse

Synonym(s):

Synaptotagmin-4, Synaptotagmin IV, SytIV

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€341.00

€341.00


Estimated to ship on11 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€341.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€341.00


Estimated to ship on11 April 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

10C11.1, monoclonal

species reactivity

human, mouse

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SYT4(6860)

General description

Synaptotagmin-4 (UniProt Q9H2B2; also known as Synaptotagmin IV, SytIV) is encoded by the SYT4 (also known as KIAA1342) gene (Gene ID 6860) in human. The mammalian synaptotagmin family is comprised of 17 members. While most of them are predominantly function in the cetnral nervous system (CNS), some are involved in the vesicular functions of endocrine cells and glucose-transport metabolic cells. Synaptotagmin-4 is an inducible member detectable only in the brain and the neuroendocrine system. Unlike other family members, synaptotagmin-4 does not bind calcium and does not medidate calcium-induced exocytosis. Instead, synaptotagmin-4 is shown to play a neuroendocrine role in inhibiting exocytotic activities in the posterior pituitary and in cultured PC12 cells. Synaptotagmin-4 is expressed most abundantly in oxytocin neurons of the hypothalamus, where it negatively regulates oxytocin exocytosis. Dietary obesity is associated with increased vesicle binding of synaptotagmin-4 and decreased oxytocin release. Normalizing oxytocin release via Synaptotagmin-4 inhibition prevents against dietary obesity under chronic nutritional excess. Human synaptotagmin-4 consists of a vesiclular domain (a.a. 1-16), a transmembrane segment (a.a. 17-37),and a large cytoplasmic domain (a.a. 38-425).

Specificity

Clone 10C11.1 targets a cytoplasmic epitope present in both human spliced isoforms reported by UniProt (Q9H2B2).

Immunogen

Epitope: Cytoplasmic domain.
GST-tagged recombinant human Synaptotagmin-4 cytoplasmic domain fragment.

Application

Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected Synaptotagmin-4 in human kidney, skeletal muscle, and pancreas tissue sections.
Research Category
Neuroscience
Research Sub Category
Vesicular Trafficking
This Anti-Synaptotagmin-4 Antibody, clone 10C11.1 is validated for use in Western Blotting, Immunohistochemistry (Paraffin) for the detection of Synaptotagmin-4.

Quality

Evaluated by Western Blotting in mouse cerebellum tissue lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected Synaptotagmin-4 in 10 µg of mouse cerebellum tissue lysate.

Target description

~48 kDa observed. 47.96/46.06 kDa (human isoform 1/2) and 47.66 kDa (mouse) calculated.

Physical form

Format: Purified
Protein G purified.
Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yazi D Ke et al.
The Journal of biological chemistry, 294(38), 14149-14162 (2019-08-02)
The microtubule-associated protein tau undergoes aberrant modification resulting in insoluble brain deposits in various neurodegenerative diseases, including frontotemporal dementia (FTD), progressive supranuclear palsy, and corticobasal degeneration. Tau aggregates can form in different cell types of the central nervous system (CNS)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service