Skip to Content
Merck
All Photos(1)

Key Documents

MABF951

Sigma-Aldrich

Anti-IL-2R beta (CD122) Antibody, clone TM-beta1

clone TM-beta 1, from rat

Synonym(s):

Interleukin-2 receptor subunit beta, IL-2 receptor subunit beta, IL-2R subunit beta, High affinity IL-2 receptor subunit beta, p70-75, CD122

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
€130.00
100 μL
€317.00

€130.00


Estimated to ship on12 May 2025Details


Request a Bulk Order

Select a Size

Change View
25 μL
€130.00
100 μL
€317.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€130.00


Estimated to ship on12 May 2025Details


Request a Bulk Order

biological source

rat

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

TM-beta 1, monoclonal

species reactivity

mouse

packaging

antibody small pack of 25 μL

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
immunoprecipitation (IP): suitable
neutralization: suitable
western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

mouse ... Il2Rb(16185)

Related Categories

General description

Interleukin-2 receptor subunit beta (UniProt: P16297; also known as IL-2 receptor subunit beta, IL-2R subunit beta, IL-2RB, High affinity IL-2 receptor subunit beta, p70-75, CD122) is encoded by the Il2rb gene (Gene ID: 16185) in murine species. IL-2R is a non-covalent dimer of an alpha and a beta subunit. It can exist as a high affinity dimer, an intermediate affinity monomer of beta subunit, and as a low affinity monomer of alpha subunit. IL-2R beta is a single-pass type I membrane protein. The beta subunit is involved in receptor mediated endocytosis and it transduces the mitogenic signals of IL-2. IL-2R beta is synthesized with a signal peptide (aa 1-26), which is cleaved off to produce the mature form that has an extracellular domain (aa 27-240), a transmembrane domain (aa 241-268), and a long cytoplasmic domain (aa 269-539). IL-2R beta contains a WSXWS motif (aa 221-225), which is essential for its proper folding and thereby efficient intracellular transport. It also has a Box 1 motif (aa 281-289) that is required for JAK interaction and/or activation.

Specificity

Clone TM-beta 1 is a rat monoclonal antibody that detects IL-2R beta subunit in murine species.

Immunogen

TART-mB whole cells

Application

Anti-IL-2R beta (CD122), clone TM-beta1, Cat. No. MABF951, is a rat monoclonal antibody that detects IL-2R beta and has been tested for use in Flow Cytometry, Function assay, Immunocytochemistry, Immunoprecipitation, Neutralizing, and Western Blotting.
Neutralizing Analysis: Freshly isolated mouse splenocytes pretreated with different amounts of MABF951, Anti-IL-2R beta (CD122), clone TM- 1 showed a co-relation of the amount of antibody used vs percent of cells stained with FITC-labeled antibody. In antibody titration, as the concentration of unlabeled anti-IL-2R beta (CD122) is decreased less inhibition is observed as a result.

Inhibits Activity/Function: A representative lot abolished the binding of IL2 ligand to IL-2R beta (CD122). (Tanaka, T., et. al. (1991). J Immunol. 147(7):2222-8; Yokoyama, S., et. al. (2009). Proc Natl Acad Sci USA. 106(37):15849-54).

Flow Cytometry Analysis: A representative lot detected IL-2R beta (CD122) in Flow Cytometry applications (Tanaka, T., et. al. (1991). J Immunol. 147(7):2222-8).

Western Blotting Analysis: A representative lot detected IL-2R beta (CD122) in Western Blotting applications (Tanaka, T., et. al. (1991). J Immunol. 147(7):2222-8).

Immunoprecipitation Analysis: A representative lot immunoprecipitated IL-2R beta (CD122) in Immunoprecipitation applications (Tanaka, T., et. al. (1991). J Immunol. 147(7):2222-8).

Immunocytochemistry Analysis: A representative lot detected IL-2R beta (CD122) in Immunocytochemistry applications (Tanaka, T., et. al. (1991). J Immunol. 147(7):2222-8).
Research Category
Inflammation & Immunology

Quality

Isotype testing: Identity Confirmation by Isotyping test.

Isotyping Analysis: The identity of this monoclonal antibody is confirmed by isotyping test to be mouse IgG2b.

Target description

60.54 kDa calculated

Physical form

Format: Purified
Protein G purified
Purified rat monoclonal antibody IgG2b in PBS without azide.

Storage and Stability

Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service