Skip to Content
Merck
All Photos(1)

Key Documents

MABF846

Sigma-Aldrich

Anti-LAG3 Antibody, clone C9B7W (Azide Free)

Synonym(s):

LAG3|Lymphocyte activation gene 3 protein|LAG-3|CD223

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€326.00

€326.00


Estimated to ship on04 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€326.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€326.00


Estimated to ship on04 April 2025Details


Request a Bulk Order

biological source

rat

Quality Level

clone

monoclonal

species reactivity

mouse

technique(s)

activity assay: suitable
flow cytometry: suitable

General description

Lymphocyte-activation gene 3 (LAG3), also known as cluster of differentiation 223 (CD223), is an immunoglobulin superfamily (IgSF) type I membrane protein encoded by the LAG3 gene in human. LAG3 is expressed in human activated T and NK cells and involved lymphocyte activation by functioning as a ligand for major histocompatibility complex (MHC) class II molecules.

Immunogen

Recombinant protein corresponding to within the extracellular D2 domain of mouse LAG3.

Application

Anti-LAG3 Antibody, clone C9B7W (Azide Free) is an antibody against LAG3 for use in Flow Cytometry, Activity Assay.
Flow Cytometry Analysis: A representative lot detected LAG3 in TILs from B16 and MC38 mice (Woo, S., et al. (2011). Cancer Research. 72(4):917-927).
Flow Cytometry Analysis: A representative lot detected LAG3 in activated T cells from CD223-/- and CD2

Quality

Evaluated by Flow Cytometry in Staph enterotoxin B treated mouse splenocyte.

Flow Cytometry Analysis: 1.0 ug of this antibody detected LAG3 in Staph enterotoxin B treated mouse splenocyte.

Other Notes

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service