Skip to Content
Merck
All Photos(1)

Key Documents

MABF406

Sigma-Aldrich

Anti-CD279 (PD-1) Antibody (mouse), PE, clone J43.1

clone J43.1, 0.2 mg/mL, from hamster(Armenian)

Synonym(s):

Programmed cell death protein 1, Protein PD-1, mPD-1, CD279

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€258.00

€258.00


Estimated to ship on22 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€258.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€258.00


Estimated to ship on22 April 2025Details


Request a Bulk Order

biological source

hamster (Armenian)

Quality Level

conjugate

PE

antibody form

purified antibody

antibody product type

primary antibodies

clone

J43.1, monoclonal

species reactivity

mouse

concentration

0.2 mg/mL

technique(s)

flow cytometry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable

isotype

IgG

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Pdcd1(18566)

General description

The J43.1 antibody is specific for mouse CD279, also known as programmed death-1 (PD-1), a 55 kDa glycoprotein which can co-regulate T cell antigen receptor signaling and therefore modulate T cell activation. PD-1 exists in a monomeric form that is expressed by CD4- CD8- thymocytes, where it participates in the processes of clonal selection, elimination of autoreactive lymphocytes, and development of tolerance. PD-1 expression is also inducible upon activation of mature T cells, where it has been proposed to interact with the co-stimulatory receptor CD80 to limit T cell activation. Two ligands for PD-1, known as PD-L1 (B7-H1) and PD-L2 (B7-DC) are differentially expressed on T and B cells, monocytes, macrophages, NK cells or dendritic cells. PD-1 is a member of a family of receptors including CD28 and CTLA-4 (CD152), which interact with “B7” ligands to provide a balance of co-stimulatory /co-inhibitory signaling important in T cell activation, tolerance, and autoimmunity.The J43.1 antibody may be used as a marker for PD-1 expression, and is commonly used for analysis of receptor-ligand interaction and function(s) in vitro and in vivo.

Immunogen

Immunogen information for this antibody is proprietary information.

Application

Research Category
Inflammation & Immunology
This armenian hamster monoclonal antibody Anti-CD279 (PD-1) (mouse), PE, clone J43.1 validated for use in IP, IHC & Flow Cytometry for the detection of CD279 (PD-1).

Quality

The monoclonal antibody was purified from tissue culture supernatant via affinity chromatography. The purified antibody was conjugated under optimal conditions, with unreacted dye removed from the preparation.

Flow Cytometry Analysis: 0.5 µg of this antibody detected CD279 (PD-1) in one million C57Bl/6 splenocytes.

Physical form

Purified Armenian Hamster Monoclonal Antibody in buffer containing 10 mM NaH2PO4, 150 mM NaCl, 0.1% gelatin, pH 7.2, and up to 0.1% sodium azide.
Size Exclusion Chromatography

Storage and Stability

It is recommended to store the product undiluted at 2-8°C and protected from prolonged exposure to light. Do not freeze.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service