Skip to Content
Merck
All Photos(1)

Key Documents

MABF3094

Sigma-Aldrich

Anti-BAFF/CD257 Antibody, clone 5A8

Synonym(s):

B-cell-activating factor, BAFF, CD257, Tumor necrosis factor ligand superfamily member 13B

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
€137.00
100 μL
€347.00

€137.00


Estimated to ship on04 April 2025Details


Request a Bulk Order

Select a Size

Change View
25 μL
€137.00
100 μL
€347.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€137.00


Estimated to ship on04 April 2025Details


Request a Bulk Order

biological source

rat

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

5A8, monoclonal

species reactivity

mouse

packaging

antibody small pack of 100

technique(s)

activity assay: suitable
direct ELISA: suitable
western blot: suitable

isotype

IgG1κ

epitope sequence

Unknown

Protein ID accession no.

UniProt accession no.

Gene Information

mouse ... Klrk1(24099)

Specificity

Clone 5A8 is a rat monoclonal antibody that detects B-cell activating factor (BAFF/CD257).

Immunogen

Rat RBL cells expressing full-length mouse BAFF.

Application

Quality Control Testing

Evaluated by Western Blotting with recombinant murine BAFF protein .

Western Blotting Analysis (WB): A 1:500 dilution of this antibody detected recombinant murine BAFF/CD257 protein .

Tested Applications

Activity Assay: A representative lot activated mouse BAFF in a reporter cell assay, in primary B cells. (Kowalczyk-Quintas, C., et al. (2016). J Biol.Chem.291(38):19826-34; Vigolo, M., et al. (2018). Nat Commun.9(1):1199).

ELISA Analysis: A representative lot detected BAFF/CD257 in ELISA applications (Batten, M., et al. (2004). J Immunol. 172(2):812-22; Vigolo, M., et al. (2018). Nat Commun. 9(1):1199; Kowalczyk-Quintas, C., et al. (2016). J Biol Chem. 291(38):19826-34).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Tumor necrosis factor ligand superfamily member 13B (UniProt: Q9WU72; also known as B-cell-activating factor, BAFF, CD257) is encoded by the Tnfsf13b (also known as Baff) gene (Gene ID: 24099) in murine species. BAFF is a single-pass type II, homotrimeric, membrane glycoprotein with a cytoplasmic domain (aa 1-47), a transmembrane domain (aa 48-68), and an extracellular domain (aa 69-309). A soluble form of BAFF has also been described that is produced by the cleavage of membrane form between amino acids 126-127. It is expressed by cells of myeloid origin and by stromal cells. Two isoforms of BAFF have been described that are produced by alternative splicing. Isoform 2 is reported to inhibit the secretion and bioactivity of isoform 1. BAFF is reported to bind to TNFRSF13B/TACI and TNFRSF17/BCMA. It serves as a potent B cell survival factor and is a critical immune checkpoint during peripheral B cell maturation. Its levels are often elevated in autoimmune diseases, such as rheumatoid arthritis and systemic lupus erythematosus (SLE). Elevated levels of BAFF are also observed in subjects with non-Hodgkin s lymphoma (NHL) and it is reported to promote survival of both NHL-derived B lymphoma cells and B cell chronic lymphocytic leukemia (B-CLL) cells. (Ref.: Vigolo, M., et al. (2018). Nat. Commun. 9(1); 1199; Kowalczyk-Quintas, C., et al (2016). J. Biol. Chem. 291(38); 19826-19834; Batten, M., et al. (2004). J. Immunol. 172(2); 812-822; Gavin, AL., et al. (2003). J. Biol. Chem. 278(40); 38220-38228).

Physical form

Purified rat monoclonal antibody IgG1 in PBS without azide.

Reconstitution

0.5 mg/mL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Store at -10°C to -25°C. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service