Skip to Content
Merck
All Photos(1)

Key Documents

MABF2752

Sigma-Aldrich

Anti-HHV-8 PF-8 Antibody, clone 11D1

Synonym(s):

PF-8;KSHV ORF59

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
€139.00
100 μG
€351.00

€139.00


Estimated to ship on07 April 2025Details


Request a Bulk Order

Select a Size

Change View
25 μG
€139.00
100 μG
€351.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

€139.00


Estimated to ship on07 April 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

11D1, monoclonal

mol wt

calculated mol wt 42.41 kDa

purified by

using protein G

species reactivity

virus

packaging

antibody small pack of 100 μg

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2b

epitope sequence

Unknown

Protein ID accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

vaccinia virus ... ORF45(4961474)

General description

Human herpesvirus-8 (also known as Kaposi′s sarcoma-associated herpesvirus, HHV-8, KSHV) is etiologically associated with all forms of Kaposi sarcoma (KS) including classic, endemic, transplant-related, and AIDS-related, as well as rare neoplastic disorders. It is a double stranded DNA virus with a large genome that encodes for over 87 open reading frames (ORFs) including genes necessary for capsid, tegument, envelope, DNA replication and regulatory proteins. HHV-8 undergoes a bi-phasic lifecycle and can persist indefinitely in the infected host in a latent form during which time only a small fraction of regulatory viral proteins is expressed. The most prominent protein expressed in latently infected cells is the ORF73 gene product known as the latency-associated nuclear antigen (LANA) that functions to ensure the maintenance of the viral genome by tethering the viral episomal DNA to host cell chromosomes. The ORF59 (also known as PF-8), a phosphoprotein, is reported to be a processivity factor for HHV-8 DNA polymerase (Pol-8). It interacts with and stimulates Pol-8 DNA synthesis activity. It is an early-late gene product localized to HHV-8-infected cell nuclei. PF-8 interacts with HHV-8 DNA polymerase (Pol-8) and stimulates Pol-8 DNA synthesis activity. Amino acid residues 10-27 and 279-301 are reported to be critical for the processivity function of PF-8. Residues 10 to 27 are essential for its interaction with Pol-8 and 279-301 are required for its binding to dsDNA. Clone 11D1 specifically detects PF-8 of HHV-8 and exhibits predominant nuclear membrane fluorescence in TPA (phorbol ester)-induced, HHV-infected BCBL-1 cells. (Ref.: Strahan, RC., et al. (2017). PLoS Pathog 13(7); e1006482; Chan, SR., and Chandran, B (2000). J. Virol. 74(23); 10920-10929; Chan, SR., et al. (1998). 240: 118-126).

Specificity

Clone 11D1 is a mouse monoclonal antibody that detects PF-8 of Kaposi′s sarcoma-associated herpesvirus (HHV-8).

Immunogen

12-O-Tetradecanoylphorbol 13-acetate (TPA)-induced BCBL-1 cells.

Application

Quality Control Testing

Evaluated by Immunocytochemistry in BCBL-1 cells.

Immunocytochemistry Analysis (ICC): A 1:50 dilution of this antibody detected Kaposi s sarcoma associated herpesvirus (HHV-8) in 12-O-Tetradecanoylphorbol 13-acetate (TPA) treated (20 ng/mL; 48 h) BCBL-1 cells.

Tested Applications

Immunoprecipitation Analysis: A representative lot immunoprecipitated PF-8 of HHV-8 in Immunoprecipitation applications (Chan, S.R., et al. (1998). Virology. 240(1):118-26; Chan, S.R., et al. (2000). J Virol. 74(23):10920-9).

Western Blotting Analysis: A representative lot detected OF-8 of HHV-8 in Western Blotting applications (Chan, S.R., et al. (1998). Virology.;240(1):118-26; Chan, S.R., et al. (2000). J Virol.;74(23):10920-9).

Immunocytochemistry Analysis: A representative lot detected PF-8 of HHV-8 in Immunocytochemistry applications (Chan, S.R., et al. (1998). Virology.;240(1):118-26).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-HHV-8 PF-8, clone 11D1, Cat. No. MABF2752, is a mouse monoclonal antibody that detects PF-8 of Kaposi′s sarcoma-associated herpesvirus (HHV-8) and is tested for use in Immunocytochemistry, Immunoprecipitation, and Western Blotting.

Physical form

Purified mouse monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Recommended storage: +2°C to +8°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service