biological source
rat
Quality Level
conjugate
unconjugated
antibody form
purified antibody
antibody product type
primary antibodies
clone
20D5, monoclonal
mol wt
observed mol wt ~27.68 kDa
species reactivity
mouse
packaging
antibody small pack of 100 μL
technique(s)
flow cytometry: suitable
inhibition assay: suitable
isotype
IgG2a
UniProt accession no.
shipped in
dry ice
storage temp.
-10 to -25°C
target post-translational modification
unmodified
Gene Information
mouse ... Klrc1(16641)
Related Categories
General description
Specificity
Immunogen
Application
Evaluated by Flow Cytometry in C57BL/6 mouse splenocytes.
Flow Cytometry Analysis: 1 μg of this antibody detected NKG2A/C/E in one million C57BL/6 mouse splenocytes.
Tested Applications
Flow Cytometry Analysis: A representative lot detected NKG2A/C/E in Flow Cytometry applications (Vance, R.E., et. al. (1999). J Exp Med. 190(12):1801-12).
Inhibition Analysis: A representative lot of this antibody completely blocked all binding of Qa-1 tetramers to NK1.11 splenocytes. (Vance, R.E., et. al. (1999). J Exp Med. 190(12):1801-12).
Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Physical form
Storage and Stability
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service