Skip to Content
Merck
All Photos(1)

Key Documents

MABF2309

Sigma-Aldrich

Anti-Adeno-associated Virus 9 Antibody, clone HL2372

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€362.00

€362.00


Estimated to ship on08 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μL
€362.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€362.00


Estimated to ship on08 April 2025Details


Request a Bulk Order

antibody form

purified antibody

clone

HL2372
monoclonal

purified by

using protein G

species reactivity

mouse, virus

concentration

(Please refer to lot specific datasheet.)

technique(s)

ELISA: suitable
dot blot: suitable

isotype

IgG2a

capacity

neutralization

General description

Adeno-associated virus 9 capsids.

Specificity

Clone HL2372 is a mouse monoclonal antibody that detects adenoassociated viruses (AAV9 and AAV8). It binds only to intact capsids and not to linear viral proteins.

Application

ELISA Analysis: A representative lot detected Adeno-associated Virus 9 in ELISA applications (Tseng, Y.S., et. al. (2016). J Virol Methods. 236:105-110).

Dot Blot Analysis: A representative lot detected Adeno-associated Virus 9 in Dot Blot applications (Tseng, Y.S., et. al. (2016). J Virol Methods. 236:105-110).

Neutralizing Analysis: A representative lot neutralized Adeno-associated Virus 9 infection in HeLa cells. (Tseng, Y.S., et. al. (2016). J Virol Methods. 236:105-110).

Quality

Evaluated by Dot Blot in AAV8 and AAV9 virus like particles.

Dot Blot Analysis: This antibody detected various concentrations of AAV8 and AAV9 virus like particles.



Physical form

Purified mouse monoclonal antibody IgG2a in PBS without preservatives.

Storage and Stability

Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Virus (Adeno-associateted virus 9).

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service