Skip to Content
Merck
All Photos(1)

Key Documents

MABF2305

Sigma-Aldrich

Anti-Adeno-associated virus 8 Antibody, clone HL2381

Synonym(s):

AAV8, Adeno-associated virus serotype 8

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
€143.00
100 μL
€362.00

€143.00


Estimated to ship on07 April 2025Details


Request a Bulk Order

Select a Size

Change View
25 μL
€143.00
100 μL
€362.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€143.00


Estimated to ship on07 April 2025Details


Request a Bulk Order

biological source

mouse

clone

HL2381, monoclonal

species reactivity

virus (Adenovirus)

isotype

IgG3κ

General description

Adeno-associated virus (AAV) is a small (25-nm), non-enveloped virus that packages a linear single-stranded DNA genome. It belongs to the family Parvoviridae and productive infection by AAV occurs only in the presence of a helper virus, which could either be adenovirus or herpesvirus. After successful infection, two stages of AAV life cycle have been described - a lytic stage and a lysogenic stage. In the presence of helper virus, the lytic stage ensues and during this period, AAV undergoes productive infection with genome replication, viral gene expression, and virion production. The lysogenic stage is established in host cells in the absence of a helper virus and AAV integrates into the host genome at specific site, AAVS1 on human chromosome 19. This site is favored due to the presence of a Rep binding element. AAV genome consists of two open reading frames, Rep and Cap, flanked by two 145 base inverted terminal repeats (ITR). The ITR base pairs allow for synthesis of the complementary DNA strand. Rep and Cap are translated to produce multiple distinct proteins. The Rep gene encodes Rep78, Rep68, Rep52, Rep40 and Cap gene produces VP1, VP2, VP3 capsid proteins. The Rep78 and Rep68 proteins participate in DNA replication process via their interaction with Rep-binding element (RBE) and terminal resolution site sequences located within the ITR. The Rep52 and Rep40 proteins are involved in the generation and accumulation of single-stranded viral genomes from double-stranded replicative intermediates. Thus far eleven serotypes of AAV have been identified, which differ in their tropism or the type of cells they infect. AAV8 shown to be suitable for use hepatic cell transduction and can serve as a useful vector for studies involving the hepatic tissue. Clone HL2381 generated against the AAV8 capsid, binds to AAV8, but also displays cross-reactivity with AAV3B. It can also neutralize infection in cells. (Ref.: Russell, WC et al. (2009). J. Gen. Virol. 90(1); 1-20; Tseng, Y-S., et al. (2016). J. Virol. Methods. 236; 105-110).

Specificity

Clone HL2381 is a mouse monoclonal antibody that detects AAV8 capsid protein. Also displays cross-reactivity with AAV3B capsid protein.

Immunogen

Recombinant Adeno-associated virus serotype 8 capsid protein.

Application

Anti-Adeno-associated virus 8, clone HL2381, Cat. No. MABF2305, is a mouse monoclonal antibody that detects Adeno-associated viruses serotype 8 and has been tested for use in Dot Blot, ELISA, and Neutralizing Applications.
Dot Blot Analysis: A representative lot detected Adeno-associated virus 8 in Dot Blot applicaitons (Tseng, Y.S., et. al. (2016). J Virol Methods. 236:105-110).

Neutralizing Analysis: A representative lot neutralized Adeno-associated virus 8 infection in cells (Tseng, Y.S., et. al. (2016). J Virol Methods. 236:105-110).

ELISA Analysis: A representative lot detected Adeno-associated virus 8 in ELISA applicaitons (Tseng, Y.S., et. al. (2016). J Virol Methods. 236:105-110).

Quality

Evaluated by Dot Blot with AAV8 and AAV9 virus like particles .

Dot Blot Analysis: Various dilutions of AAV8 and AAV9 virus like particles were probed with Adenovirus 8. This antibody detects AAV8, but not AAV9.

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG3 in PBS without preservatives.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.

Other Notes

Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service