Skip to Content
Merck
All Photos(1)

Key Documents

MABF2293

Sigma-Aldrich

Anti-IL-10 Antibody, clone 2H6

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
€143.00
100 μG
€362.00

€143.00


Estimated to ship on02 April 2025Details


Request a Bulk Order

Select a Size

Change View
25 μG
€143.00
100 μG
€362.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€143.00


Estimated to ship on02 April 2025Details


Request a Bulk Order

antibody form

purified antibody

clone

2H6
monoclonal

purified by

using protein G

species reactivity

mouse, human, human

concentration

(Please refer to lot specific datasheet.)

technique(s)

affinity binding assay: suitable
immunohistochemistry: suitable

isotype

IgG1κ

UniProt accession no.

General description

Full-length recombinant mature human IL-10 (without the signal peptide).

Specificity

Clone 2H6 is a mouse monoclonal antibody that specifically detects human interleukin-10 (IL-10).

Application

Affinity binding assay: The binding of Anti-IL-10, clone 2H6, Cat. No. MABF2293, showed a KD value of 1.68 x 10-9, whereas binding to GST showed a KD of 7.22 x 10-3.

Quality

Evaluated by Immunohistochemistry (Paraffin) in human tonsil tissue sections.

Immunohistochemistry (Paraffin) Analysis: A 1:250 dilution of this antibody detected IL-10 in human tonsil tissue sections.

Physical properties

20.52 kDa calculated.

Physical form

Purified mouse monoclonal antibody IgG1 in PBS with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service