Skip to Content
Merck
All Photos(1)

Key Documents

MABF2156

Sigma-Aldrich

Anti-TLR9 Antibody, clone NaR9

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€362.00

€362.00


Estimated to ship on15 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μL
€362.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€362.00


Estimated to ship on15 April 2025Details


Request a Bulk Order

antibody form

purified antibody

clone

NaR9
monoclonal

purified by

using protein G

species reactivity

mouse, mouse, mouse

concentration

(Please refer to lot specific datasheet.)

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable

isotype

IgG2aκ

UniProt accession no.

Mode of action

| inhibits

General description

Ba/F3 cells expressing full-length mouse TLR9 protein. Immunization was carried out on Tlr9 -/- mice.

Specificity

Clone NaR9 is a mouse monoclonal antibody that specifically detects Toll-like receptor 9 in murine species. It targets an epitope within the N-terminal region.

Application

Immunocytochemistry Analysis: A representative lot detected TLR9 in Immunocytochemistry applications (Murakami, Y., et. al. (2017). Sci Rep. 7:44042).

Flow Cytometry Analysis: A representative lot detected TLR9 in Flow Cytometry applications (Murakami, Y., et. al. (2017). Sci Rep. 7:44042).

Inhibits Activity/Function: A representative lot inhibited TLR9-dependent cytokine production in vitro by bone marrow-derived macrophages and conventional dendritic cells. (Murakami, Y., et. al. (2017). Sci Rep. 7:44042).

Quality

Evaluated by Immunocytochemistry in Raw264.7 cells.

Immunocytochemistry Analysis: A 1:250 dilution of this antibody detected TLR9 in Raw264.7 cells.

Physical properties

116.41 kDa calculated.

Physical form

Purified mouse monoclonal antibody IgG2a in PBS without azide.

Storage and Stability

Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service