Skip to Content
Merck
All Photos(2)

Key Documents

MABF2055

Sigma-Aldrich

Anti-IDO-2 Antibody, clone 4-3

clone 4-3, from mouse

Synonym(s):

Indoleamine 2, 3-dioxygenase 2, Indoleamine 2, 3-dioxygenase-like protein 1, Indoleamine-pyrrole 2, 3-dioxygenase-like protein 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€330.00

€330.00


Estimated to ship on04 June 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€330.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€330.00


Estimated to ship on04 June 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4-3, monoclonal

species reactivity

mouse

packaging

antibody small pack of 25 μg

technique(s)

western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

mouse ... Ido2(209176)

General description

Indoleamine 2,3-dioxygenase 2 (UniProt: Q8R0V5; also known as IDO-2, Indoleamine 2,3-dioxygenase-like protein 1, Indoleamine-pyrrole 2,3-dioxygenase-like protein 1) is encoded by the Ido2 (also known as Indol1) gene (Gene ID: 209176) in murine species. IDO-2 is expressed mainly in antigen-presenting immune cells, liver, kidney, brain, and placenta. It is highly expressed in kidney, followed by epididymis and liver and is also detected in the tails of the spermatozoa. It catalyzes the first and rate-limiting step in the kynurenine pathway of tryptophan catabolism. IDO-1 and IDO-2 are 2 distinct enzymes which catalyze the same reaction. However, the Km for IDO-2 for tryptophan is much higher than that of IDO-1. IDO-2 may play a role as a negative regulator of IDO-1 by competing for heme-binding with IDO-1. IDO-2 activity is inhibited by 1MT (1-methyl-tryptophan) and MTH-trp (methylthiohydantoin-DL-tryptophan). IDO-2 is also involved in immune regulation. IDO-2 knockout mice do not show any apparent defects in their embryonic development or hematopoietic differentiation and have wild-type profiles for kynurenine in blood serum and for immune cells in spleen, lymph nodes, peritoneum, thymus and bone marrow. However, the knockout mice exhibit defects in IDO-mediated T-cell regulation and inflammatory responses. (Ref.: Mbongue JC, et al. (2015). Vaccines (Basel). 3(3):703-29).

Specificity

Clone 4-3 detects IDO-2 in mouse. It targets an epitope within 21 amino acids in the C-terminal region.

Immunogen

KLH-conjugated linear peptide/recombinant protein corresponding to 21 amino acids from the C-terminal region of murine Indoleamine 2,3-dioxygenase 2 (IDO-2).

Application

Anti-IDO-2, clone 4-3, Cat. No. MABF2055, is a mouse monoclonal antibody that detects IDO2 and has been tested for use in Western Blotting.
Research Category
Inflammation & Immunology
Western Blotting Analysis: 0.5 µg/mL from a represesntative lot detected IDO-2 in 10 µg of mouse kidney tissue lysate.

Quality

Evaluated by Western Blotting in mouse liver tissue lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected IDO-2 in 10 µg of mouse liver tissue lysate.

Target description

~55 kDa observed; 45.25 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service