Skip to Content
Merck
All Photos(1)

Key Documents

MABF1824

Sigma-Aldrich

Mouse IgG1 Negative Control, clone MOPC-21, redFluor® 710 conjugate Antibody

clone MOPC-21, from mouse

Synonym(s):

Immunoglobulin G1, IgG1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€188.00

€188.00


Estimated to ship on10 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€188.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€188.00


Estimated to ship on10 April 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

clone

MOPC-21, monoclonal

technique(s)

flow cytometry: suitable

isotype

IgG1

shipped in

wet ice

target post-translational modification

unmodified

General description

The MOPC-21 immunoglobulin is useful as an isotype-matched control. The MOPC-21 immunolglobulin has an unknown binding specificity and is used as an isotype control for mouse IgG1 antibodies.

Application

This mouse monoclonal Mouse IgG1 Negative Control, clone MOPC-21, redFluor® 710 conjugate, Cat. No. MABF1824 is validated for use in Flow Cytometry.

Quality

This monoclonal antibody was purified from tissue culture supernatant via affinity chromatography.

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Legal Information

redFluor is a registered trademark of Tonbo Biotechnologies

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service